CellLight® MAP4-GFP, BacMam 2.0 - FAQs
CellLight® MAP4-GFP, BacMam 2.0 - FAQs
View additional product information for CellLight® MAP4-GFP, BacMam 2.0 - FAQs (C10598)
2 product FAQs found
Do you offer an alternative for the discontinued product CellLight MAP4-GFP (Cat. No. C10598)?
No. Unfortunately, we currently do not have a direct replacement available for the discontinued product CellLight MAP4-GFP (Cat. No. C10598) in our portfolio. However, we do offer MAP4 antibodies as an alternative. It's important to note that these antibodies are not specifically indicated for live cell imaging.
I keep getting low transduction efficiency when using CellLight labeling reagents on my neurons. What can I do to improve the efficiency?
Neurons are more difficult to transduce than many other cells. The main way to improve transduction is to label with a higher number of particles per cell. For primary neurons, it can also help to transduce them at the time of plating rather than on established cultures. There can also be a slower onset of expression in neurons and peak expression often occurs on day 2-3 rather than 16 hours after transduction.