CellLight® MAP4-RFP, BacMam 2.0 - FAQs
CellLight® MAP4-RFP, BacMam 2.0 - FAQs
View additional product information for CellLight® MAP4-RFP, BacMam 2.0 - FAQs (C10599)
1 product FAQs found
I keep getting low transduction efficiency when using CellLight labeling reagents on my neurons. What can I do to improve the efficiency?
Neurons are more difficult to transduce than many other cells. The main way to improve transduction is to label with a higher number of particles per cell. For primary neurons, it can also help to transduce them at the time of plating rather than on established cultures. There can also be a slower onset of expression in neurons and peak expression often occurs on day 2-3 rather than 16 hours after transduction.