Cell Sorting Set-up Beads (for UV lasers) - FAQs
Cell Sorting Set-up Beads (for UV lasers) - FAQs
View additional product information for Cell Sorting Set-up Beads (for UV lasers) - FAQs (C16506)
1 product FAQs found
What are Cell Sorting Set-up Beads used for?
Cell Sorting Set-up Beads (Cat. Nos. C16506, C16507, C16508, and C16509) are reliable standards for flow cytometry cell sorter instrument setup and calibration. The Cell Sorting Set-up Beads approximate the size, emission wavelength, and intensity of many biological samples, and so can be used to calibrate a flow cytometer's cell sorting system, including laser source, optics, and stream flow. The beads are optimized for checking cell sorter settings such as drop delay and efficiency (cell loss during sorting). Cell Sorting Set-up Beads are available for use with UV, blue, green/yellow, and red lasers.