Product Image
Invitrogen™

SMD1168, Pichia pastoris Yeast Strain

Several Pichia pastoris strains are available to allow you to optimize expression and recovery of your protein of interest. TableRead more
Have Questions?
Catalog NumberQuantity
C175001 stab
Catalog number C17500
Price (USD)
648.00
Each
Quantity:
1 stab
Price (USD)
648.00
Each
Several Pichia pastoris strains are available to allow you to optimize expression and recovery of your protein of interest. Table 1 provides information to help you choose the appropriate strain.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Bacterial or Yeast StrainSMD1168
For Use With (Application)Yeast Expression
Product TypeYeast Strain
Quantity1 stab
Cell TypeYeast
SpeciesP. pastoris
Unit SizeEach

Frequently asked questions (FAQs)

When selecting for blasticidin-resistant transformants in the X-33 strain using pPIC6/pPIC6α vectors, why do I get large and small colonies on YPD plates containing 300 µg/ml blasticidin?

Generally, large colonies represent transformants containing pPIC6/pPIC6α integrants, while small colonies represent transformants containing pPIC6/pPIC6α non-integrants. These non-integrants have transduced the pPIC6/pPIC6α plasmid, and therefore, exhibit a low level of blasticidin resistance in the initial selection process. Upon subsequent screening, these non-integrant transformants do not retain blasticidin resistance.

When choosing a blasticidin-resistant transformant for your expression studies, we recommend that you pick blasticidin-resistant colonies from the initial transformation plate and streak them on a second YPD plate containing the appropriate concentration of blasticidin. Select transformants that remain blasticidin-resistant for further studies.

Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.

My transformation is not working. Do you have any suggestions?

Here are some suggestinos:

- Make sure that you have harvested cells during log-phase growth (OD <1.0 generally).
- If electroporation is being used, see the electroporator manual for suggested conditions. Vary electroporation parameters if necessary.
- Use more DNA.
- Use freshly made competent cells.
- If the LiCl transformation method is being used, try boiling the carrier DNA.

Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.

My spheroplasting of Pichia worked twice, but hasn't worked since. The OD of the culture simply does not drop.

Here are some things to consider:

- If the OD of cells that are used is too high, they will not spheroplast. Do not overgrow cells.
- Do not use old cells and make sure that they are in log phase of growth.
- Make sure to mix zymolyase well before using. Zymolyase is more of a suspension than a solution.
- Make the PEG solution fresh each time and check the pH.

Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.

What are the different kinds of media used for culturing Pichia pastoris and S. cerevisiae?

Following are the rich and minimal media used for culturing Pichia pastoris and S. cerevisiae:

Rich Media:
S. cerevisiae and Pichia pastoris
YPD (YEPD): yeast extract, peptone, and dextrose
YPDS: yeast extract, peptone, dextrose, and sorbitol

Pichia pastoris only
BMGY: buffered glycerol-complex medium
BMMY: buffered methanol-complex medium

Minimal Media (also known as drop-out media):
S. cerevisiae
SC (SD): Synthetic complete (YNB, dextrose (or raffinose or galactose), and amino acids)

Pichia pastoris
MGY: minimal glycerol medium
MD: minimal dextrose
MM: minimal methanol
BMGH: buffered minimal glycerol
BMMH: buffered minimal methanol

Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.

Is there a recommended protocol for fermentation using constitutive expression vectors such as pGAPZ?

Use the following high cell density protocol for pGAP clones. Feed carbon until the desired density is reached (300 to 400 g/L wet cell weight (WCW)). If the protein is well-behaved in the fermenter, increase to 300-400 g/L WCW as with methanol inducible clones. These densities can be reached in less than 48 hours of fermentation. We have fermented constitutive expressers on glycerol using these protocols with good results. Some modifications to the Fermentation Basal Salts Medium that you might want to make are:

1) Substitute 2% dextrose for the 4% glycerol in the batch medium.
2) Substitute 40% dextrose for the 50% glycerol in the fed-batch medium.
3) Feed the 40% dextrose at 12 mL/L/hr (Jim Cregg has published data on expression using several carbon sources as substrates; dextrose gave the highest levels of expression).
4) Yeast extract and peptone may be added to the medium for protein stability.

One warning: If you are working with His- strains, they remain His- after transformation with pGAPZ. Fermentation in minimal medium will require addition of histidine to the fermenter.

Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.

Citations & References (5)

Citations & References
Abstract
The fifth epidermal growth factor-like domain of thrombomodulin does not have an epidermal growth factor-like disulfide bonding pattern.
Authors:White CE, Hunter MJ, Meininger DP, Garrod S, Komives EA
Journal:Proc Natl Acad Sci U S A
PubMed ID:8816772
'The disulfide bonding pattern of the fourth and fifth epidermal growth factor (EGF)-like domains within the smallest active fragment of thrombomodulin have been determined. In previous work, this fragment was expressed and purified to homogeneity, and its cofactor activity, as measured by Kcat for thrombin activation of protein C, was ... More
Production of human tissue factor using the Pichia pastoris expression system.
Authors:Austin AJ, Jones CE, Heeke GV
Journal:Protein Expr Purif
PubMed ID:9631526
Tissue factor plays an important role in the initiation of the blood coagulation cascade resulting in the formation of a fibrin clot. The extracellular domain of human tissue factor has been expressed in the protease-deficient strain of the methylotrophic yeast Pichia pastoris, SMD1168. Tissue factor was expressed with a human ... More
Identification and characterization of two alternative splice variants of human interleukin-2.
Authors:Tsytsikov VN, Yurovsky VV, Atamas SP, Alms WJ, White B
Journal:J Biol Chem
PubMed ID:8798495
Our previous work showed that alternative splicing is used to make an inhibitory variant of human interleukin (IL)-4. Because of homology between IL-4 and IL-2 proteins and receptors, we tested whether alternative splicing is used to generate similar inhibitory variants of human IL-2. Messenger RNA from peripheral blood mononuclear cells ... More
Laccase from the white-rot fungus Trametes versicolor: cDNA cloning of lcc1 and expression in Pichia pastoris.
Authors:Jonsson LJ, Saloheimo M, Penttila M
Journal:Curr Genet
PubMed ID:9388299
A cDNA coding for laccase was isolated from the ligninolytic fungus Trametes versicolor by RNA-PCR. The cDNA corresponds to the gene lcc1, which encodes a laccase isoenzyme of 498 amino-acid residues preceded by a 22-residue signal peptide. The lcc1 cDNA was cloned into the vector pHIL-D2 for expression in Pichia ... More
The donor substrate specificity of the human beta 1,3-glucuronosyltransferase I toward UDP-glucuronic acid is determined by two crucial histidine and arginine residues.
Authors: Ouzzine Mohamed; Gulberti Sandrine; Levoin Nicolas; Netter Patrick; Magdalou Jacques; Fournel-Gigleux Sylvie;
Journal:J Biol Chem
PubMed ID:11986319
The human beta1,3-glucuronosyltransferase I (GlcAT-I) plays a key role in proteoglycan biosynthesis by catalyzing the transfer of glucuronic acid onto the trisaccharide-protein linkage structure Galbeta1,3Galbeta1,4Xylbeta-O-Ser, a prerequisite step for polymerization of glycosaminoglycan chains. In this study, we identified His(308) and Arg(277) residues as essential determinants for the donor substrate (UDP-glucuronic ... More