Owl™ C2-S Micro Electrophoresis System
Owl™ C2-S Micro Electrophoresis System
Owl™ C2-S Micro Electrophoresis System
Thermo Scientific™

Owl™ C2-S Micro Electrophoresis System

Screen up to seven samples on one gel in less than five minutes with the Thermo Scientific™ Owl™ C2-S Micro Electrophoresis System.
Have Questions?
Catalog NumberGel Size
C2-S5 x 7.5 cm
Catalog number C2-S
Price (USD)
432.00
Online exclusive
480.00
Save 48.00
Each
Gel Size:
5 x 7.5 cm
Price (USD)
432.00
Online exclusive
480.00
Save 48.00
Each
Screen up to seven samples on one gel in less than five minutes with the Thermo Scientific™ Owl™ C2-S Micro Electrophoresis System.
Features:
  • Horizontal microgel system is designed for quick restriction fragment analysis.
  • Microgel may also be used to quickly check a few PCR samples before loading a large number of samples on a larger gel, saving time
  • Glass slides, 2 × 3 in. (5 × 7.5cm), serve as the gel tray
  • To use, pour and form thin gels by surface tension directly on glass slide; complete run lasts five to 10 minutes
  • Using available caster and casting slides, gels can be cast and solidified in less than 10 minutes.
Includes:
  • Buffer chamber (buffer volume, 100mL)
  • Lid
  • Power supply leads
  • Casting tray
  • 10 casting slides
  • One seven-well comb, 1.5mm thick
Specifications
DescriptionHorizontal Electrophoresis System; Gel size: 5 x 7.5cm
Dimensions (L x W x H)6.75 x 2.63 x 4.38 in. (17 x 38 x 11 cm)
Height (English)4.38 in.
Height (Metric)11 cm
Length (English)6.75 in.
Length (Metric)17 cm
Length (Metric) Gel7.5 cm
Sample Size7
Volume (Metric) Buffer100 mL
Width (English)2.63 in.
Width (English) Gel6.5
Width (Metric)38 cm
Width (Metric) Gel5 cm
Gel Size5 x 7.5 cm
Product LineOwl
TypeMicro
Unit SizeEach

Frequently asked questions (FAQs)

Are Bio-Rad power supplies compatible with Owl Electrophoresis systems?

The Owl Electrophoresis Systems are supplied with standard power leads, but we have not tested compatibility of the Owl systems with the Bio-Rad power supplies. For best performance, we recommend using one of our Owl or PowerEase power supplies for the Owl Electrophoresis Systems.