Owl™ C2-S Micro Electrophoresis System Buffer Chamber and Lid, 100mL, 17 x 6.5 x 6.5cm (L x W x H) - FAQs
Owl™ C2-S Micro Electrophoresis System Buffer Chamber and Lid, 100mL, 17 x 6.5 x 6.5cm (L x W x H) - FAQs
View additional product information for Owl™ C2-S Micro Electrophoresis System Replacement Parts - FAQs (C2)
1 product FAQs found
Are Bio-Rad power supplies compatible with Owl Electrophoresis systems?
The Owl Electrophoresis Systems are supplied with standard power leads, but we have not tested compatibility of the Owl systems with the Bio-Rad power supplies. For best performance, we recommend using one of our Owl or PowerEase power supplies for the Owl Electrophoresis Systems.