Do you have ethnicity information for your human epidermal melanocyte donor lines?
We do provide age/sex/pigmentation/ethnicity on the Certificate of Analysis (COA).
Search
View additional product information for Human Epidermal Melanocytes, neonatal, darkly pigmented donor, (HEMn-DP), Proliferating Culture, Prepared in Medium 254 and HMGS-2 - FAQs (C20225PB)
1 product FAQs found