Product Image
Invitrogen™
CandyCane™ Glycoprotein Molecular Weight Standards
CandyCane glycoprotein molecular weight standards separate into alternating bands of glycosylated and nonglycosylated proteins during polyacrylamide gel electrophoresis.Read more
Have Questions?
Catalog NumberQuantity
C21852
also known as C-21852
400 μL
Catalog number C21852
also known as C-21852
Price (USD)
399.00
Each
Quantity:
400 μL
Request bulk or custom format
Price (USD)
399.00
Each
CandyCane glycoprotein molecular weight standards separate into alternating bands of glycosylated and nonglycosylated proteins during polyacrylamide gel electrophoresis.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Gel CompatibilityNovex™ Tricine Gels, Novex™ Tris-Glycine Gels, NuPAGE™ Bis-Tris Gels, NuPAGE™ Tris-Acetate Gels, SDS-PAGE Gels
Quantity400 μL
Ready to LoadNo
Shipping ConditionRoom Temperature
Product LineCandyCane
Product TypeGlycoprotein Molecular Weight Standard
Size Range14 to 180 kDa
Stain TypeUnstained
System TypeSDS-PAGE
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C).
Frequently asked questions (FAQs)
I used one of your protein standards for a western transfer and noticed that some of the lower-molecular weight protein bands passed through the membrane. How can I resolve this issue?
- Decrease voltage, current or length of transfer time
- Make sure that the methanol concentration in the transfer buffer is proper; use a methanol concentration of 10-20% methanol removes the SDS from SDS-protein complexes and improves the binding of protein to the membrane.
- Make sure that the SDS concentration (if added) in the transfer buffer is proper, don't use more than 0.02-0.04% SDS. Using too much SDS can prevent binding of proteins to the membrane.
- Check the pore size of the membrane and the size of the target protein. Proteins smaller than 10 kDa will easily pass through a 0.45 µm pore size membrane. If proteins smaller than 10 kDa are of interest, it would be better to use a 0.2 µm pore size membrane.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
I used one of your protein standards for a western transfer and noticed that some of the higher-molecular weight bands transferred very poorly to the membrane. Can you offer some tips?
- Increase voltage, current or length of time for transfer
- SDS in the gel and in the SDS-protein complexes promotes elution of the protein from the gels but inhibits binding of the protein to membranes. This inhibition is higher for nitrocellulose than for PVDF. For proteins that are difficult to elute from the gel such as large molecular weight proteins, a small amount of SDS may be added to the transfer buffer to improve transfer. We recommend pre-equilibrating the gel in 2X Transfer buffer (without methanol) containing 0.02-0.04% SDS for 10 minutes before assembling the sandwich and then transferring using 1X transfer buffer containing 10% methanol and 0.01%SDS.
- Methanol removes the SDS from SDS-protein complexes and improves the binding of protein to the membrane, but has some negative effects on the gel itself, leading to a decrease in transfer efficiency. It may cause a reduction in pore size, precipitation of some proteins, and some basic proteins to become positively charged or neutral. Make sure that the methanol concentration in the transfer buffer is not more than 10-20% and that high-quality, analytical grade methanol is used.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
I used one of your pre-stained standards on a Tris-Glycine gel and noticed that the molecular weights of the proteins were different than on a NuPAGE Bis-Tris gel. What is the reason for this?
Pre-stained standards have a dye that is covalently bound to each protein that will result in the standard migrating differently in different buffer systems (i.e., different gels). As a result, using a pre-stained standard for molecular weight estimation will only give the apparent molecular weight of the protein. Pre-stained standards may be used for molecular weight approximation, confirming gel migration and estimating blotting efficiency but for accurate molecular weight estimation, an unstained standard should be used.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
I used one of your protein standards and am seeing some extra bands in the lane. Can you offer some suggestions?
- While loading, take care to make sure that there is no cross-contamination from adjacent sample lanes.
- Make sure that the correct amount of standard is loaded per lane. Loading too much protein can result in extra bands and this is a problem especially with silver-stained gels.
- Improper storage of the standard or repeated freeze/thawing can result in protein degradation.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
I used one of your protein standards and the bands look non-distinct and smeary. What should I do?
Here are some suggestions:
- Make sure that the correct amount of standard is loaded per lane. Loading too much protein can cause smearing and this is a problem especially with silver stained gels.
- Bands will not be as well resolved in low percentage gels. Try using a higher percentage gel.
- If the bands look smeary and non-distinct after a western transfer/detection, this may be due to the antibody being too concentrated. Follow the manufacturer's recommended dilution or determine the optimal antibody concentration by dot-blotting.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
Citations & References (9)
Citations & References
Abstract
Simultaneous red/green dual fluorescence detection on electroblots using BODIPY TR-X succinimidyl ester and ELF 39 phosphate.
Authors:Martin K, Hart C, Schulenberg B, Jones L, Patton WF
Journal:Proteomics
PubMed ID:11987124
'A two-color fluorescence detection method is described based upon covalently coupling the succinimidyl ester of BODIPY TR-X dye to proteins immobilized on polyvinylidene difluoride membranes, followed by detection of target proteins using the fluorogenic, precipitating substrate ELF 39-phosphate in combination with alkaline phosphatase conjugated reporter molecules. This results in all ... More
Myofibrillar Ca(2+) sensitivity is uncoupled from troponin I phosphorylation in hypertrophic obstructive cardiomyopathy due to abnormal troponin T.
Authors:Bayliss CR, Jacques AM, Leung MC, Ward DG, Redwood CS, Gallon CE, Copeland O, McKenna WJ, Dos Remedios C, Marston SB, Messer AE,
Journal:Cardiovasc Res
PubMed ID:23097574
'We studied the relationship between myofilament Ca(2+) sensitivity and troponin I (TnI) phosphorylation by protein kinase A at serines 22/23 in human heart troponin isolated from donor hearts and from myectomy samples from patients with hypertrophic obstructive cardiomyopathy (HOCM). We used a quantitative in vitro motility assay. With donor heart ... More
Ehrlichia chaffeensis expresses macrophage- and tick cell-specific 28-kilodalton outer membrane proteins.
Authors:Singu V, Liu H, Cheng C, Ganta RR
Journal:Infect Immun
PubMed ID:15618143
'Ehrlichia chaffeensis, a tick-transmitted rickettsial agent, causes human monocyte/macrophage-tropic ehrlichiosis. In this study, proteomic approaches were used to demonstrate host cell-specific antigenic expression by E. chaffeensis. The differentially expressed antigens include those from the 28-kDa outer membrane protein (p28-Omp) multigene locus. The proteins expressed in infected macrophages are the products ... More
GtfA and GtfB are both required for protein O-glycosylation in Lactobacillus plantarum.
Authors:Lee IC, van Swam II, Tomita S, Morsomme P, Rolain T, Hols P, Kleerebezem M, Bron PA,
Journal:
PubMed ID:24532775
Acm2, the major autolysin of Lactobacillus plantarum WCFS1, was recently found to be O-glycosylated with N-acetylhexosamine, likely N-acetylglucosamine (GlcNAc). In this study, we set out to identify the glycosylation machinery by employing a comparative genomics approach to identify Gtf1 homologues, which are involved in fimbria-associated protein 1 (Fap1) glycosylation in ... More
Shiga-like toxin binds with high avidity to multivalent O-linked blood group P1 determinants on mucin-type fusion proteins.
Authors:Maria Cherian R, Gaunitz S, Nilsson A, Liu J, Karlsson NG, Holgersson J,
Journal:
PubMed ID:24082034
The binding of Shiga-like toxin 1 (Stx1) and Shiga-like toxin 2 (Stx2) to a mucin-like fusion protein, P-selectin glycoprotein ligand-1/mouse IgG2b (PSGL-1/mIgG2b), carrying multiple copies of the blood group P1 determinant on O-glycans was investigated with western blot and the biosensor Biacore. Chinese hamster ovary K-1 (CHO-K1) cells were stably ... More