CellTrace™ Blue Cell Proliferation Kit, for flow cytometry
Invitrogen™

CellTrace™ Blue Cell Proliferation Kit, for flow cytometry

The CellTrace Blue Cell Proliferation Kit is used for in vitro and in vivo labeling of cells to trace multipleRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
C3457420 Assays
C34568180 Assays
Catalog number C34574
Price (USD)
144.00
Each
Quantity:
20 Assays
Price (USD)
144.00
Each

The CellTrace Blue Cell Proliferation Kit is used for in vitro and in vivo labeling of cells to trace multiple generations using dye dilution by flow cytometry. The CellTrace Blue reagent is specifically designed for excitation by a UV laser (at 355 nm or 375 nm), and it can be easily multiplexed with reagents and antibodies excited by other commonly used lasers.

• Superior performance—bright, single-peak staining enables visualization of multiple generations
• Long-term signal stability—well-retained in cells for several days post stain
• Versatile—multiple colors available to easily combine with antibodies or markers of cell function, such as GFP
• Simple, robust staining protocol

Superior fluorescent staining
Successful proliferation analysis by dye dilution (see figure below) requires an extremely bright dye to distinguish fluorescently labeled cells from auto-fluorescence after several cell divisions. The intense fluorescent staining provided by CellTrace Blue dye enables the visualization of five or more generations of proliferating cells before the signal is overwhelmed by intrinsic cellular auto-fluorescence. Consistent, homogeneous staining results in very little fluorescence variation between cells in a population, so distinct generations can be seen without any requirement for complex modeling software.The recommended working concentration for peripheral blood mononuclear cells is 5–10 μM. However, as with all reagents used in flow cytometry, to achieve optimal signal, reagent titration is highly recommended.

Long-term signal retention
Unlike stains that label the lipid membrane of cells, CellTrace Blue stain easily crosses the plasma membrane and covalently binds inside cells where the stable, well-retained fluorescent dye provides a consistent signal, even after several days in a cell culture environment. CellTrace Blue dye binds covalently to all free amines on the surface and inside of cells and shows little cytotoxicity, with minimal observed effect on the proliferative ability or biology of cells.

Easy multiplexing with other fluorophores
The UV excitation and narrow emission of CellTrace Blue dye make it ideal for multiplexing as it lies outside the spectral emission profiles of most other dyes and fluorescent proteins.

Simple, robust staining protocol
The CellTrace Blue Cell Proliferation Kit contains convenient single-use vials of dry dye to permit small-scale experiments without preparing excess quantities of dye. A stock solution is prepared by dissolving the contents of a vial in anhydrous DMSO prior to use. To stain 1 mL of cells in protein-free medium, 1 μL of this stock solution is typically used. Cells should be stained for 20 minutes at room temperature with gentle agitation. A brief wash with complete medium will then quench any dye remaining in solution.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodFluorescence
Dye TypeCellTrace™Blue
Emission Wavelength355/410
Quantity20 Assays
Shipping ConditionRoom Temperature
For Use With (Application)Proliferation Assay
For Use With (Equipment)Flow Cytometer
Product LineCellTrace
Product TypeBlue Cell Proliferation Kit
Unit SizeEach
Contents & Storage
  • 1 vial lyophilized CellTrace Blue dye
  • 100 μL DMSO
  • Store in freezer -5°C to -30°C.

Frequently asked questions (FAQs)

I would like to label two cell populations with two different CellTrace reagents and then co-culture these cells. Will the CellTrace reagent leave the cells to stain other cells?

We have not tested the use of the CellTrace reagents for co-culture applications. In theory, this may work, but you would have to test this on your cells of interest.

Find additional tips, troubleshooting help, and resources within our Cell Tracing and Tracking Support Center.

With the CellTrace cell proliferation kits, for flow cytomtery, I am getting only one broad peak on my histogram instead of multiple peaks. What is causing this?

A single broad peak is usually caused by using too high a concentration of dye and/or too long an incubation time.

Find additional tips, troubleshooting help, and resources within our Flow Cytometry Support Center.

What cellular components do the CellTrace Proliferation dyes label?

The dyes provided in the CellTrace Proliferation kits are amine-reactive dyes that are considered general cytoplasmic stains. They may bind to various membrane proteins, organelles as well as components in the cytoplasm. They are not known to localize in any specific organelles.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.