CholEsteryl 4,4-Difluoro-5,7-Dimethyl-4-Bora-3a,4a-Diaza-s-Indacene-3-Dodecanoate (CholEsteryl BODIPY™ FL C12) - FAQs
CholEsteryl 4,4-Difluoro-5,7-Dimethyl-4-Bora-3a,4a-Diaza-s-Indacene-3-Dodecanoate (CholEsteryl BODIPY™ FL C12) - FAQs
View additional product information for CholEsteryl 4,4-Difluoro-5,7-Dimethyl-4-Bora-3a,4a-Diaza-s-Indacene-3-Dodecanoate (CholEsteryl BODIPY™ FL C12) - FAQs (C3927MP)
3 product FAQs found
How should I store CholEsteryl 4,4-Difluoro-5,7-Dimethyl-4-Bora-3a,4a-Diaza-s-Indacene-3-Dodecanoate (CholEsteryl BODIPY FL C12) (Cat. No. C3927MP) once reconstituted?
Once reconstituted in either chloroform or alcohol, store frozen, desiccated and protected from light. The CholEsteryl BODIPY FL C12 is not sensitive to changes in temperature (freeze/thaw), but the gradual accumulation of water from condensation and from the atmosphere will promote aggregation of the reagent over time.
Find additional tips, troubleshooting help, and resources within our Cell Imaging Support Center.
What is the molecular weight of CholEsteryl 4,4-Difluoro-5,7-Dimethyl-4-Bora-3a,4a-Diaza-s-Indacene-3-Dodecanoate (CholEsteryl BODIPY FL C12) (Cat. No. C3927MP)? Is the molecular weight based on the cholesteryl moiety together with BODIPY?
The molecular weight of the product is 786.98 g/mol. This is the molecular weight of the full molecule.
Find additional tips, troubleshooting help, and resources within our Cell Imaging Support Center.
How do I reconstitute CholEsteryl 4,4-Difluoro-5,7-Dimethyl-4-Bora-3a,4a-Diaza-s-Indacene-3-Dodecanoate (CholEsteryl BODIPY FL C12) (Cat. No. C3927MP)?
CholEsteryl BODIPY FL C12 is soluble up to 1 mM in chloroform. The molecular weight of this product is 786.98 g/mol. Additionally, there are references to the product being soluble up to 25 mM in ethanol and up to 0.5 mg/mL in methanol with sonication.
Find additional tips, troubleshooting help, and resources within our Cell Imaging Support Center.