8mm Assembled Clear Autosampler Vial Kits
Actual product may vary
Thermo Scientific™
8mm Assembled Clear Autosampler Vial Kits
Convenience kits save time and reduce the risk of contamination. Each convenience kit includes 100 vials pre-assembled with 100 screw thread caps and septa.
Have Questions?
Catalog NumberClosure ColorClosure TypeSeptumPackaging Type
C4015-100YellowScrew Cap, Open TopIntegrated MembraneStorage Tray
Catalog number C4015-100
Price (USD)
-
Closure Color:
Yellow
Closure Type:
Screw Cap, Open Top
Septum:
Integrated Membrane
Packaging Type:
Storage Tray
Save time during sample preparation and reduce the risk of contamination by using Thermo Scientific™ 8mm Clear Vial Convenience Kits. Each kit includes 100 clear vials pre-assembled with 100 screw thread caps and septa. They are packaged in convenient vial trays with clear covers or in economical polybags..
  • Packaged in convenient vial trays with clear covers or in economical polybags
  • Vial trays are stackable for easy storage
  • All caps are manufactured from virgin polypropylene unless noted
Includes:
100 vials with pre-attached caps and septa
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Closure ColorYellow
Closure MaterialPolypropylene
Closure Size8-425
DescriptionClear Polypropylene (PP) Microvial, Yellow Polypropylene (PP) Cap, Integrated Membrane
Diameter (Metric) Hole5.5 mm
Packaging TypeStorage Tray
Septa Size (Metric)8-425 mm
SeptumIntegrated Membrane
Closure TypeScrew Cap, Open Top
ColorClear
Marking SpotNo
MaterialPolypropylene
Pre-slitNo
Volume (Metric)2 mL
Unit SizePack of