13 mm Autosampler Vial Screw Thread Caps
Thermo Scientific™

13 mm Autosampler Vial Screw Thread Caps

13mm Screw Thread Caps feature a 13-425 thread finish. Available in an assortment of styles, pre-assembled caps and septa minimize contamination.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorSeptumMaterialProduct Type
C4015-30ABlack, Ivory, RedIvory PTFE/Red RubberPTFE, Rubber, RubberOpen Top Screw Cap
C4015-1ABlackNonePolypropyleneOpen Top Screw Cap
C4015-66ABlack, Red, WhiteSilicone/PTFEPTFE, PTFE, SiliconeOpen Top Screw Cap
C4015-40ABlack, Red, WhitePTFE/Silicone/PTFEPTFE, PTFE, SiliconeOpen Top Screw Cap
C4015-55BLKBlack, Blue, WhiteSilicone/PTFEPTFE, Silicone, SiliconeOpen Top Screw Cap
C4015-5ANaturalPolyethylene, IntegralPolyethylene, PolyethyleneScrew Cap
12-SCR
also known as 12-SC(R)
RedNonePolypropyleneOpen Top Screw Cap
12-SCY
also known as 12-SC(Y)
YellowNonePolypropyleneOpen Top Screw Cap
Catalog number C4015-30A
Price (USD)
54.51
Pack of 100
Color:
Black, Ivory, Red
Septum:
Ivory PTFE/Red Rubber
Material:
PTFE, Rubber, Rubber
Product Type:
Open Top Screw Cap
Price (USD)
54.51
Pack of 100
Thermo Scientific™ 13mm Screw Thread Caps feature a 13-425 thread finish and are available in an assortment of styles to match your HPLC, GC or IC application needs. Convenient pre-assembled caps and septa minimize contamination from handling. Choose from Thermo Scientific™, Thermo Scientific™ National™, and Thermo Scientific™ Chromacol™ brands.
  • Open-top
  • 13-425 GPI Thread
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
AutoclavableAutoclavable
Certifications/ComplianceSpecification Certified
Closure Size13 mm
Color (Seal)Ivory/Red
DescriptionPP; Black; Ivory PTFE/Red Rubber septum
DisposableSingle-use
For Use With (Equipment)13-425 Screw Thread Vials
Product TypeOpen Top Screw Cap
SeptumIvory PTFE/Red Rubber
Thickness (Metric)1.5 mm
ColorBlack, Ivory, Red
MaterialPTFE, Rubber, Rubber
Quantity100/Pk.
SterilityNon-sterile
Unit SizePack of 100