MultiShot™ FlexPlate TOP10 Competent Cells
Invitrogen™

MultiShot™ FlexPlate TOP10 Competent Cells

MultiShot FlexPlate TOP10 chemically competent E. coli cells are cloning competent cells that are pre-aliquoted in a 96-well PCR plateRead more
Have Questions?
Catalog NumberQuantity
C4081201
also known as C40812-01
1 x 96 well plate (20 μl/well)
Catalog number C4081201
also known as C40812-01
Price (USD)
-
Quantity:
1 x 96 well plate (20 μl/well)

MultiShot FlexPlate TOP10 chemically competent E. coli cells are cloning competent cells that are pre-aliquoted in a 96-well PCR plate to increase transformation productivity. The FlexPlate format can be divided down to any combination of 8-well segments to allow for flexibility in transformation throughput.

Versatile TOP10 cloning capabilities
MultiShot FlexPlate TOP10 Competent Cells are the laboratory workhorse for chemical transformations as they are compatible with a broad spectrum of DNA types for transformation. The TOP10 strain is ideal for high-efficiency cloning and plasmid propagation, achieving transformation efficiency of >1 x 108 cfu/μg with control plasmid DNA in FlexPlate format. It allows stable replication of high-copy number plasmids and is the same competent cell strain that come with many of our cloning kits.

TOP10 E. coli cells are genetically similar to the DH10B strain, but has been reported to be more resilient to stress conditions (osmotic shock and acidic pH stress). These cells carry mutations in the methylation-dependent restriction system (mcrA, mcrBC, and mrr), allowing the cloning of both prokaryotic and eukaryotic genomic DNA, as well as efficient plasmid rescue from eukaryotic genomes. In addition, recA1 and endA1 mutations increase insert stability and improve the quality of plasmid DNA prepared from minipreps.

MultiShot FlexPlate TOP10 Competent Cells offer:
• Transformation efficiencies of >1 x 108 cfu/μg
• hsdR for efficient transformation of unmethylated DNA from PCR amplifications
• mcrA for efficient transformation of methylated DNA from genomic preparations
• lacZΔM15 for blue/white color screening of recombinant clones on plates containing either X-Gal or Bluo-Gal
• endA1 for cleaner preparations of DNA and better results in downstream applications due to the elimination of non-specific digestion by endonuclease I
• recA1 for reduced occurrence of non-specific recombination in cloned DNA

Note: TOP10 cells do not require IPTG to induce expression from the lac promoter.

Increase transformation productivity with MultiShot FlexPlate format
The FlexPlate is a 96-well PCR plate that can be used in automated liquid handling systems or divided into 8-well segments for flexibility in transformation throughput. Addition of DNA to the MultiShot FlexPlate Competent Cells can be quickly performed with a multichannel pipet or a liquid handling system. After addition of DNA, the cells can be efficiently transformed by heat-shock using a water bath, heat block, or thermal cycler.

Key benefits of the MultiShot FlexPlate format
• Increased throughput—compatible with automated liquid handling platforms
• Flexible—transform in increments of 8 wells to entire 96-well plate
• Convenient—competent cells pre-aliquoted into 96-well plate, ready for transformation

Genotype
F–mcrA Δ(mrr-hsdRMS-mcrBC) φ80lacZΔM15 ΔlacX74 recA1 araD139 Δ(ara-leu)7697 galU galK λ–rpsL(StrR) endA1 nupG

Find the strain and format that you need
We offer many other E. coli strains and formats of chemically competent cells and electrocompetent cells to help meet specific transformation needs. For high-throughput transformation, choose from our collection of MultiShot formatted comp cells or email us for custom formatting options. Bacterial growth media is also available in various formats.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Antibiotic Resistance BacterialYes (Streptomycin)
Blue/White ScreeningYes (lacZΔM15)
Cloning Methylated DNAYes (mcrA)
Cloning Unstable DNANo
Contains F' EpisomeNo
High-throughput CompatibilityHigh
Improves Plasmid QualityYes (endA1)
PlasmidHigh Copy Plasmid
Preparing Unmethylated DNANo
Product LineMultishot
Product TypeChemically Competent Cells
Quantity1 x 96 well plate (20 μl/well)
Reduces RecombinationYes (recA1)
Shipping ConditionDry Ice
T1 Phage - Resistant (tonA)No
Format96-well Plate
SpeciesE. coli (K12)
Unit SizeEach
Contents & Storage
• MultiShot FlexPlate TOP10 Competent Cells (1 x 96-well plate at 20 μL/well)
Store at –80°C.

• pUC19 DNA (50 μL at 10 pg/μL)
Store at –20°C.

• S.O.C. Medium (2 x 10 mL)
Store at 4°C or room temperature.

Frequently asked questions (FAQs)

Is it possible to perform a fast transformation protocol with the MultiShot competent cells?

There are situations where either because of time or process limitations, elimination of the heat shock and recovery time post-heat shock in the transformation protocol would be desirable. Testing was performed on many of our chemically competent cells in both the MultiShot StripWell and MultiShot FlexPlate format with a shortened transformation protocol. Use of a rapid transformation protocol (add DNA, no heat shock, no recovery period, plate on warm agar) yielded up to a log lower in transformation efficiency using the control DNA. This rapid transformation protocol can be performed with expectation of lower colony counts than our standard high efficiency transformation protocol and can only be performed with plasmids that use ampicillin as their antibiotic resistance. The rapid protocol is ideal when using a liquid handling system for high-throughput transformations with high cloning efficiency. Please see the product manual for full details on this rapid transformation protocol option.

Can I use a thermal cycler or heat block to heat shock the competent cells in the MultiShot FlexPlate?

The MultiShot FlexPlate is a PCR plate and can be placed in a thermal cycler or heat block set at 42 degrees C to transform the cells by the heat shock method. The duration of the heat shock performed in these devices has yielded similar transformation efficiencies as that seen with heat shock done in a water bath.

Which E. coli strains do you offer in the MultiShot StripWell format?

We offer the following E. coli strains in the MultiShot StripWell format: TOP10, DH5alpha T1R, Mach1 T1R, Stbl3, and BL21 Star (DE3) chemically competent cells.

Which E. coli strains do you offer in the MultiShot FlexPlate format?

We offer the following E. coli strains in the MultiShot FlexPlate format: TOP10, DH5alpha T1R, DH10B T1R, Mach1 T1R, OmniMAX2 T1R, and Stbl3 chemically competent cells.

What are the differences between the MultiShot StripWell, MultiShot FlexPlate, and MultiShot formats of E. coli cells?

The MultiShot StripWell format is supplied as pre-aliquoted competent cells in 96 tubes contained in a rack. The tubes are attached in strips of 8 tubes, and there are 12 strips that contain 50 µL of chemically competent cells. This format offers the most flexibility to transform 1 tube or up to all 96 tubes at once and is intended for varied or medium-throughput use.

The MultiShot FlexPlate format is supplied as pre-aliquoted competent cells in a 96-well PCR plate. Each well contains 20 µL of competent cells. The plate can be separated at each column of 8 wells, offering a high level of flexibility to utilize various increments of 8 wells per transformation. The wells of the plate are heat sealed, preventing detachment of the aluminum seal in the -80 degrees C freezer and preventing contamination. This format is compatible with automated liquid handling platforms and is intended for medium- to high-throughput use.

The MultiShot format is supplied as pre-aliquoted competent cells in 5, 96-well thin-wall polycarbonate microplates. Each well of the 96-well microplate contains 15 µL of chemically competent cells. This format is compatible with automated liquid handling platforms and is intended for high-throughput use.