Are there any special considerations when using Zeocin for selection in E. coli?
Also, anyE. coli strain that contains the complete Tn5 transposable element (i.e. DH5?F'IQ, SURE, SURE2) encodes the ble (bleomycin) resistance gene. These strains can confer resistance to Zeocin. For the most efficient selection, use anE. coli strain that does not contain the Tn5 gene (i.e. TOP10, DH5a, DH10B, etc.).