MultiShot&trade; StripWell TOP10 Chemically Competent <i>E. coli</i>
Invitrogen™

MultiShot™ StripWell TOP10 Chemically Competent E. coli

MultiShot StripWell TOP10 Chemically Competent E. coli cells are ideal for high-efficiency cloning and plasmid propagation, achieving a transformation efficiencyRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
C409601
also known as C4096-01
1 x 96 tube rack (50 μl/tube)
C409605
also known as C4096-05
5 x 96 tube racks (50 μl/tube)
C409610
also known as C4096-10
10 x 96 tube racks (50 μl/tube)
Catalog number C409601
also known as C4096-01
Price (USD)
1,015.65
Online exclusive
1,080.00
Save 64.35
Each
Quantity:
1 x 96 tube rack (50 μl/tube)
Price (USD)
1,015.65
Online exclusive
1,080.00
Save 64.35
Each

MultiShot StripWell TOP10 Chemically Competent E. coli cells are ideal for high-efficiency cloning and plasmid propagation, achieving a transformation efficiency of >1 x 108 cfu/μg with control plasmid DNA in StripWell format. These cloning competent cells are packaged in a rack containing 12 strips of 8 tubes to increase productivity for throughput bacterial transformations.

Versatile TOP10 cloning capabilities 
MultiShot StripWell TOP10 Chemically Competent E. coli cells are the laboratory workhorse for chemical transformations as they are compatible with a broad spectrum of DNA types for transformation. They allow stable replication of high-copy number plasmids and are the same competent cells that come with many of our cloning kits.

TOP10 E. coli cells are genetically similar to the DH10B strain and have been reported to be more resilient to stress conditions (osmotic shock and acidic pH stress). These cells carry mutations in the methylation-dependent restriction system (mcrA, mcrBC, and mrr), allowing cloning of both prokaryotic and eukaryotic genomic DNA, as well as efficient plasmid rescue from eukaryotic genomes. Similar to other DH strains, TOP10 cells contain recA1 and endA1 mutations that increase insert stability and improve the quality of plasmid DNA prepared from minipreps.

MultiShot StripWell TOP10 Chemically Competent E. coli cells offer:
• Transformation efficiencies of >1 x 108 cfu/μg
• hsdR for efficient transformation of unmethylated DNA from PCR amplifications
• mcrA for efficient transformation of methylated DNA from genomic preparations
• lacZΔM15 for blue/white color screening of recombinant clones on plates containing either X-Gal or Bluo-Gal
• endA1 for cleaner preparations of DNA and better results in downstream applications due to the elimination of non-specific digestion by endonuclease I
• recA1 for reduced occurrence of non-specific recombination in cloned DNA

Note: TOP10 cells do not require IPTG to induce expression from the lac promoter.

MultiShot StripWell flexible format
Each StripWell of eight connected tubes is designed with the same single-tube, single-use features as our One Shot format tubes. This format allows all steps of the transformation protocol, up to plating, to take place in the same tube, thereby saving time and reducing chance of contamination. After addition of DNA, MultiShot StripWell competent cells can be transformed by heat-shock at 42°C using a water bath, all in the same tube.

StripWell tubes can be cut to permit transformations in a single tube, eliminating wasted aliquots and avoiding freeze-thaws that can result in reduced transformation efficiency.

Benefits of the MultiShot StripWell format include:
• Increased productivity—suitable for high throughput experimentation
• Maximal flexibility—transform as few as 1 tube or as many as 96 from the StripWell rack
• Familiar convenience—add DNA directly to cells, heat shock, and recover in the same tube

Genotype
F–mcrA Δ(mrr-hsdRMS-mcrBC) φ80lacZΔM15 ΔlacX74 recA1 araD139 Δ(ara-leu)7697 galU galK λ–rpsL(StrR) endA1 nupG

Find the strain and format that fits your needs
Many other E. coli strains and formats of chemically competent and electrocompetent cells are also available to meet specific transformation needs.
For high-throughput transformation, choose from our collection of MultiShot formatted comp cells or email us for custom formatting options.
We also offer bacterial growth media in various formats.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Antibiotic Resistance BacterialYes (Streptomycin)
Blue/White ScreeningYes (lacZΔM15)
Cloning Methylated DNAYes (mcrA)
Cloning Unstable DNANo
Contains F' EpisomeNo
High-throughput CompatibilityHigh
Improves Plasmid QualityYes (endA1)
PlasmidHigh Copy Plasmid
Preparing Unmethylated DNANo
Product LineMultiShot
Product TypeChemically Competent Cells
Quantity1 x 96 tube rack (50 μl/tube)
Reduces RecombinationYes (recA1)
Shipping ConditionDry Ice
T1 Phage - Resistant (tonA)No
Transformation Efficiency LevelMedium Efficiency (1 x 108 to 1 x 109 cfu/μg)
Format8-well Strips
SpeciesE. coli (K12)
Unit SizeEach
Contents & Storage
• MultiShot StripWell TOP10 Chemically Competent E. coli (1 x 96-stripwell plate at 50 μL/well)
Store Competent Cells at –80°C.

• pUC19 DNA (1 x 50 μL vial at 10 pg/μL)
Store pUC19 DNA at –20°C.

• S.O.C. medium (2 x 15 mL bottles)
Store S.O.C. Medium at 4°C or room temperature.

Frequently asked questions (FAQs)

Is it possible to perform a fast transformation protocol with the MultiShot competent cells?

There are situations where either because of time or process limitations, elimination of the heat shock and recovery time post-heat shock in the transformation protocol would be desirable. Testing was performed on many of our chemically competent cells in both the MultiShot StripWell and MultiShot FlexPlate format with a shortened transformation protocol. Use of a rapid transformation protocol (add DNA, no heat shock, no recovery period, plate on warm agar) yielded up to a log lower in transformation efficiency using the control DNA. This rapid transformation protocol can be performed with expectation of lower colony counts than our standard high efficiency transformation protocol and can only be performed with plasmids that use ampicillin as their antibiotic resistance. The rapid protocol is ideal when using a liquid handling system for high-throughput transformations with high cloning efficiency. Please see the product manual for full details on this rapid transformation protocol option.

Can I use a thermal cycler or heat block to heat shock the competent cells in the MultiShot FlexPlate?

The MultiShot FlexPlate is a PCR plate and can be placed in a thermal cycler or heat block set at 42 degrees C to transform the cells by the heat shock method. The duration of the heat shock performed in these devices has yielded similar transformation efficiencies as that seen with heat shock done in a water bath.

Which E. coli strains do you offer in the MultiShot StripWell format?

We offer the following E. coli strains in the MultiShot StripWell format: TOP10, DH5alpha T1R, Mach1 T1R, Stbl3, and BL21 Star (DE3) chemically competent cells.

Which E. coli strains do you offer in the MultiShot FlexPlate format?

We offer the following E. coli strains in the MultiShot FlexPlate format: TOP10, DH5alpha T1R, DH10B T1R, Mach1 T1R, OmniMAX2 T1R, and Stbl3 chemically competent cells.

What are the differences between the MultiShot StripWell, MultiShot FlexPlate, and MultiShot formats of E. coli cells?

The MultiShot StripWell format is supplied as pre-aliquoted competent cells in 96 tubes contained in a rack. The tubes are attached in strips of 8 tubes, and there are 12 strips that contain 50 µL of chemically competent cells. This format offers the most flexibility to transform 1 tube or up to all 96 tubes at once and is intended for varied or medium-throughput use.

The MultiShot FlexPlate format is supplied as pre-aliquoted competent cells in a 96-well PCR plate. Each well contains 20 µL of competent cells. The plate can be separated at each column of 8 wells, offering a high level of flexibility to utilize various increments of 8 wells per transformation. The wells of the plate are heat sealed, preventing detachment of the aluminum seal in the -80 degrees C freezer and preventing contamination. This format is compatible with automated liquid handling platforms and is intended for medium- to high-throughput use.

The MultiShot format is supplied as pre-aliquoted competent cells in 5, 96-well thin-wall polycarbonate microplates. Each well of the 96-well microplate contains 15 µL of chemically competent cells. This format is compatible with automated liquid handling platforms and is intended for high-throughput use.