I need to load liposomes with calcein. Should I use the AM form or the non-AM form?
Calcein, AM requires esterase cleavage of the acetoxymethyl (AM) ester to become fluorescent. Liposomes don't have esterases unless specifically constructed to include the enzyme. The water-soluble, non-AM form of calcein (Cat. No. C481), does not require esterase cleavage to be fluorescent.