One Shot&trade; BL21(DE3)pLysE Chemically Competent <i>E. coli</i>
Invitrogen™
One Shot™ BL21(DE3)pLysE Chemically Competent E. coli
One Shot BL21(DE3)pLysE Chemically Competent E. coli cells are ideal for use with bacteriophage T7 promoter-based expression systems such asRead more
Have Questions?
Catalog NumberQuantity
C65650321 x 50 μL
Catalog number C656503
Price (USD)
434.65
Online exclusive
457.00
Save 22.35
Each
Quantity:
21 x 50 μL
Price (USD)
434.65
Online exclusive
457.00
Save 22.35
Each
One Shot BL21(DE3)pLysE Chemically Competent E. coli cells are ideal for use with bacteriophage T7 promoter-based expression systems such as pRSET and pET. The pLysE plasmid carried by the BL21(DE3)pLysE strain produces higher amounts of T7 lysozyme than pLysS to further reduce basal level expression of the gene of interest. Like pLysS, pLysE confers resistance to chloramphenicol (CamR) and contains the p15A origin for compatibility with plasmids containing the ColE1 or pMB1 origin (i.e. pUC- or pBR322- derived plasmids). Recombinant proteins that are nontoxic to E. coli are generally expressed at higher levels in BL21(DE3) cells as compared to the BL21(DE3)pLysE strain with tighter expression control. The BL21(DE3)pLysE strain is recommended for use when expressing toxic genes (alternatively, BL21-AI could be considered).
The BL21 (DE3) E. coli strain is one of the most popular host strains to produce recombinant proteins and adapted in research or industrial applications (e.g., first biopharmaceutical recombinant human insulin). This strain had been engineered to carry the λDE3 lysogen that contains the T7 RNA polymerase gene under control of the lacUV5 promoter. IPTG is required to induce expression of the T7 RNA polymerase cascade system that in turn transcribe the T7 promoter regulated target gene.
BL21(DE3) is a derivative of the E. coli B strain that does not contain the lon protease and is also deficient in the outer membrane protease OmpT. The lack of two key proteases reduces degradation of heterologous proteins expressed in the strains. Additionally, several other strain features make them especially suited for protein production, namely fast growth in minimal medium, a lower acetate production when grown in high glucose media, and the ability to reach high cell density.
One Shot BL21(DE3)pLysE Chemically Competent E. coli offer:
• Transformation efficiency of >1 x 107 cfu/μg control plasmid DNA
• Ion and OmpT protease deficiency reduces degradation of recombinant protein
• IPTG-inducible lacUV5 promoter controls expression of the T7 RNA polymerase
• pLysE (CamR) plasmid for reduced basal expression in uninduced cells
• hsdS mutation allows efficient transformation of unmethylated DNA
• Fast growth in minimal medium and ability to reach high cell density
BL21(DE3)pLysE E. coli are ideal for use with T7 promoter-based expression vectors. Like BL21(DE3)pLysS E. coli, BL21(DE3)pLysE cells carry the lambda DE3 lysogen. In addition, BL21(DE3)pLysE cells contain the pLysE plasmid, which constitutively expresses T7 lysozyme. T7 lysozyme reduces the basal expression of target genes by inhibiting T7 RNA polymerase. The pLysE plasmid in BL21(DE3)pLysE expresses higher levels of T7 lysozyme than the pLysS plasmid in BL21(DE3)pLysS. The BL21(DE3)pLysE strain provides tighter control of T7 RNA polymerase, which is necessary when the recombinant protein to be expressed is toxic.
Easy-to-use One Shot format
BL21(DE3)pLysE Chemically Competent E. coli cells are supplied in the convenient, single-reaction One Shot format. The single-tube, single-use format allows all steps of the transformation protocol, up to plating, to take place in the same tube, thereby helping save time and prevent contamination.
Genotype
F–ompT hsdSB (rB–, mB–) gal dcm (DE3) pLysE(CamR)
Find the strain and format that you need
We offer other E. coli strains for protein expression.
For expression of toxic proteins, consider BL21-AI One Shot Chemically Competent E. coli.
BL21 Star (DE3) and other strains are available in MultiShot formats for high throughput applications.
Explore bacterial growth media formats.
We offer a variety of systems for the expression of recombinant proteins in E. coli, the Champion pET expression system provides the highest level of protein production available in any expression system.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Antibiotic Resistance BacterialYes (Chloramphenicol)
Blue/White ScreeningNo
Cloning Methylated DNANo
Contains F' EpisomeNo
High-throughput CompatibilityLow
Improves Plasmid QualityNo
Improves Protein StabilityYes (lon, ompT)
Improves RNA StabilityYes (pLysE)
Preparing Unmethylated DNAYes (dcm)
Product LineOne Shot
Product TypeChemically Competent Cells
Quantity21 x 50 μL
Reduces RecombinationNo
Shipping ConditionDry Ice
T1 Phage - Resistant (tonA)No
Toxic ProteinsNo
Transformation Efficiency LevelSubcloning Efficiency (1 x 106 to 1 x 107 cfu/μg)
FormatTube
PromoterT7
SpeciesE. coli (B)
Unit SizeEach
Contents & Storage
• BL21 Star (DE3)pLysE Chemically Competent E. coli (21 x 50 μL); store at –80°C
• pUC19 DNA (1 x 50 μL); store at –20°C
• S.O.C. Medium (6 mL); store at 4°C or room temperature
Frequently asked questions (FAQs)
My gene of interest is toxic to bacterial cells. Are there any precautions you can suggest?
Several precautions may be taken to prevent problems resulting from basal level expression of a toxic gene of interest. These methods all assume that the T7-based or Champion-based expression plasmid has been correctly designed and created.
- Propagate and maintain your expression plasmid in a strain that does not contain T7 RNA polymerase (i.e., DH5α).
- If using BL21 (DE3) cells, try growing cells at room temperature rather than 37 degrees C for 24-48 hr.
- Perform a fresh transformation using a tightly regulated E. coli strain, such as BL21-AI cells.
- After following the transformation protocol, plate the transformation reaction on LB plates containing 100 µg/mL ampicillin and 0.1% glucose. The presence of glucose represses basal expression of T7 RNA polymerase.
- Following transformation of BL21-AI cells, pick 3 or 4 transformants and inoculate directly into fresh LB medium containing 100 µg/mL ampicillin or 50 µg/mL carbenicillin (and 0.1% glucose, if desired). When the culture reaches an OD600 of 0.4, induce expression of the recombinant protein by adding L-arabinose to a final concentration of 0.2%.
- When performing expression experiments, supplement the growth medium with 0.1% glucose in addition to 0.2% arabinose.
- Try a regulated bacterial expression system such as our pBAD system.
Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.
I'm trying to express my protein using a bacterial expression system. How do I know if I'm seeing degradation of my protein or if what I’m seeing is codon usage bias?
Typically, if you see 1-2 dominant bands, translation stopped prematurely due to codon usage bias. With degradation, you usually see a ladder of bands. With degradation, you can try using a protease inhibitor and add it to the lysis buffer to help prevent degradation. If degradation is the issue, a time point experiment can be done to determine the best time to harvest the cells.
Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.
I'm trying to express my protein using a bacterial expression system and am getting inclusion bodies. What should I do?
If you are having a solubility issue, try to decrease the temperature or decrease the amount of IPTG used for induction. You can also try a different, more stringent cell strain for expression. Adding 1% glucose to the bacterial culture medium during expression can also help.
Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.
I'm getting low protein yield from my bacterial expression system. What can I do to improve this?
- Inoculate from fresh bacterial cultures, since higher protein yields are generally obtained from a fresh bacterial colony.
- Check the codon usage in the recombinant protein sequence for infrequently used codons. Replacing the rare codons with more commonly used codons can significantly increase expression levels. For example, the arginine codons AGG and AGA are used infrequently by E. coli, so the level of tRNAs for these codons is low.
- Add protease inhibitors, such as PMSF, to buffers during protein purification. Use freshly made PMSF, since PMSF loses effectiveness within 30 min of dilution into an aqueous solution.
- If you are using ampicillin for selection in your expression experiments, you may be experiencing plasmid instability due to the absence of selective conditions. This occurs as the ampicillin is destroyed by β-lactamase or hydrolyzed under the acidic media conditions generated by bacterial metabolism. You may want to substitute carbenicillin for ampicillin in your transformation and expression experiments.
- The recombinant protein may be toxic to bacterial cells. Try a tighter regulation system for competent cell expression such as BL21-AI. You may also consider trying a different expression system such as the pBAD system.
Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.
My cells are growing very slowly, and I'm not getting any protein expression from my baterial expression system. What can I do to fix this?
This typically occurs when your gene of interest is toxic. Try using a tighter regulation system, such as BL21 (DE3) (pLysS) or BL21 (DE3) (pLysE), or BL21(AI).
Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.
Citations & References (1)
Citations & References
Abstract
A novel group of oleosins is present inside the pollen of Arabidopsis.
Authors: Kim Hyun Uk; Hsieh Kai; Ratnayake Chandra; Huang Anthony H C;
Journal:J Biol Chem
PubMed ID:11929861
In plants, subcellular triacylglycerol granules in seeds (oil bodies) and floral tapetum (tapetosomes) are stabilized by amphipathic structural protein called oleosin. We hereby report a novel group of oleosins that is present inside the pollen of Arabidopsis thaliana. We have used the conserved sequence of oleosins to locate, via the ... More