I just found out that Library Efficiency DB3.1 and One Shot ccdB Survival T1R cells have been discontinued. Do you offer an alternative for propagating ccdB-containing plasmids?
We offer One Shot ccdB Survival 2 T1R cells (Cat. No. A10460) for propagating ccdB-containing plasmids.