Nalgene™ PC Ultra-Low Particulate Biotainers
El producto real puede variar
Thermo Scientific™

Nalgene™ PC Ultra-Low Particulate Biotainers

Designed for the most critical applications, Nalgene PC Ultra-Low Particulate Biotainers are platinum certified clean to contain less than one-third of the USP <788> particulate limits. With exceptionally low particulates and endotoxins, they set the standard for biological and pharmaceutical manufacturing downstream applications.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCapacidad (métrico)Forma
CE-N3030-42125 mLCuadrado
CE-N3500-2020 mLRedondo
CE-N3120-421 LCuadrado
CE-N3233-422 LCuadrado
CE-N3405-425 LCuadrado
CE-N3410-4210 LCuadrado
Número de catálogo CE-N3030-42
Precio (USD)
-
Capacidad (métrico):
125 mL
Forma:
Cuadrado
The Thermo Scientific™ Nalgene™ multi-tiered portfolio provides bioproduction facilities with a variety of high-quality, sterile storage and transport solutions. This enables you to choose the appropriate product necessary to suit your application—through the step-down process and into your clean room.

Multiple tiers

  • Platinum certified clean containers are certified to contain less than 1/3 of the allowable particulate limits specified in USP <788>
  • Certified clean containers meet USP <788> low particulate requirements

Multiple resins available

  • Polycarbonate (PC)
  • Polyethylene terephthalate (PETG)
  • High-density polyethylene (HDPE)

Sterile, ready-to-use products

  • Robust triple-layer packaging for your step-down process
    • 2 heat-sealed bags create dual sterile barriers
    • 1 carton liner
    • Double-walled cartons
    • Improved labeling
  • Produced in clean room (ISO class 7)
  • Certified leakproof* to protect valuable contents
Especificaciones
Esterilizables en autoclaveNot Autoclavable
Revestimiento del tapónLined
Material del cierrePolycarbonate
Tamaño del tapón38 mm
Tipo de tapónCap
Diámetro (métrico)52.1 mm
Para utilizar con (aplicación)Media/Buffer Preparation ​, Media/Buffer Storage, Sampling, Storage, Transportation ​, Validation Testing, Drug Substance, Drug Product
Altura (imperial)4.13 in.
Altura (métrico)104.6 mm
Niveles bajos de partículas<1/3 USP <788> (Ultralow particulate)
Intervalo de temperatura (métrico)-100°C to 100°C
Dispensación a través del tapónNo
Anchura (imperial)12.5 in.
Paso del flujo de trabajoDownstream validation, downstream intermediate storage and transport. Bulk drug product/substance transportation
Capacidad (imperial)4,23 oz
Capacidad (métrico)125 mL
Certificaciones/conformidadUSP Class VI
Closure IncludedSí
ColorClear
GraduatedSí
MaterialPolycarbonate
BocaNarrow
Tipo de productoStorage and Transport Bottle
FormaCuadrado
EsterilidadEstéril
Unit SizeEach