Specifications
AutoclavableNot Autoclavable
Capacity (English)33.8 oz.
Certifications/ComplianceUSP Class VI, USP 661, FDA 21 CFR 177.1315
Closure LinerLined
Closure MaterialPolypropylene
Closure Size48 mm
Closure TypeCap
Diameter (Metric)98 mm
For Use With (Application)Media/Buffer Preparation , Media/Buffer Storage, Sampling, Storage, Transportation , Validation Testing, Drug Substance, Drug Product
Height (English)7.91 in.
Height (Metric)201 mm
Liner MaterialSilicone
Product TypeStorage and Transport Bottle
ShapeSquare
Temperature Range (Metric)-100°C to 100°C
Through the Closure DispensingNo
Width (English)20.5 in.
Workflow StepDownstream validation, downstream intermediate storage and transport. Bulk drug product/substance transportation
Capacity (Metric)1 L
Closure IncludedYes
ColorClear
GraduatedYes
MaterialPolycarbonate
MouthNarrow
SterilitySterile
Unit SizeCase of 35