Nalgene™ PC Ultra-Low Particulate Biotainers
Actual product may vary
Thermo Scientific™

Nalgene™ PC Ultra-Low Particulate Biotainers

Designed for the most critical applications, Nalgene PC Ultra-Low Particulate Biotainers are platinum certified clean to contain less than one-third of the USP <788> particulate limits. With exceptionally low particulates and endotoxins, they set the standard for biological and pharmaceutical manufacturing downstream applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Shape
CE-N3405-425 LSquare
CE-N3500-2020 mLRound
CE-N3030-42125 mLSquare
CE-N3120-421 LSquare
CE-N3233-422 LSquare
CE-N3410-4210 LSquare
Catalog number CE-N3405-42
Price (USD)
-
Capacity (Metric):
5 L
Shape:
Square
The Thermo Scientific™ Nalgene™ multi-tiered portfolio provides bioproduction facilities with a variety of high-quality, sterile storage and transport solutions. This enables you to choose the appropriate product necessary to suit your application—through the step-down process and into your clean room.

Multiple tiers

  • Platinum certified clean containers are certified to contain less than 1/3 of the allowable particulate limits specified in USP <788>
  • Certified clean containers meet USP <788> low particulate requirements

Multiple resins available

  • Polycarbonate (PC)
  • Polyethylene terephthalate (PETG)
  • High-density polyethylene (HDPE)

Sterile, ready-to-use products

  • Robust triple-layer packaging for your step-down process
    • 2 heat-sealed bags create dual sterile barriers
    • 1 carton liner
    • Double-walled cartons
    • Improved labeling
  • Produced in clean room (ISO class 7)
  • Certified leakproof* to protect valuable contents
Specifications
AutoclavableNot Autoclavable
Closure LinerLined
Closure MaterialPolypropylene
Closure Size48 mm
Closure TypeCap
Diameter (Metric)166 mm
For Use With (Application)Media/Buffer Preparation ​, Media/Buffer Storage, Sampling, Storage, Transportation ​, Validation Testing, Drug Substance, Drug Product
Height (English)11.77 in.
Height (Metric)299 mm
Liner MaterialSilicone
Temperature Range (Metric)-100°C to 100°C
Through the Closure DispensingNo
Width (English)14.3 in.
Workflow StepDownstream validation, downstream intermediate storage and transport. Bulk drug product/substance transportation
Capacity (English)169 oz.
Capacity (Metric)5 L
Certifications/ComplianceUSP
Closure IncludedYes
ColorClear
GraduatedYes
MaterialPolycarbonate
MouthNarrow
Product TypeStorage and Transport Bottle
ShapeSquare
SterilitySterile
Unit SizeCase of 6