TurboFlow™ Columns for Automated Online LC-MS Sample Cleanup
Actual product may vary
Thermo Scientific™
TurboFlow™ Columns for Automated Online LC-MS Sample Cleanup
Green features
Automate online matrix cleanup of complex matrices on your LC-MS to eliminate manual cleanup and improve productivity and certainty of results.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionColumn PhaseDiameter (Metric) Outer
CH-953285TurboFlow Cyclone C2 ColumnSilica based reversed phase0.5 mm
CH-952434TurboFlow Cyclone ColumnPolymer based1 mm
CH-952813TurboFlow Cyclone MCX ColumnPolymer based phase mixed cation exchange1 mm
CH-952979TurboFlow Cyclone MAX ColumnPolymer based mixed mode anion exchange1 mm
CH-953244TurboFlow Cyclone C18 ColumnSilica based reversed phase1 mm
CH-953275TurboFlow Cyclone C18-P ColumnSilica based reversed phase1 mm
CH-953276TurboFlow Cyclone C8 ColumnSilica based reversed phase1 mm
CH-953277TurboFlow Cyclone Fluro ColumnSilica based fluorinated alkyl1 mm
CH-953279TurboFlow Cyclone C2 ColumnSilica based reversed phase1 mm
CH-953280TurboFlow Cyclone C18 ColumnSilica based reversed phase0.5 mm
CH-953281TurboFlow Cyclone C18-P ColumnSilica based reversed phase0.5 mm
CH-953282TurboFlow Cyclone C8 ColumnSilica based reversed phase0.5 mm
CH-953283TurboFlow Cyclone Fluro ColumnSilica based fluorinated alkyl0.5 mm
CH-953286TurboFlow Cyclone MAX ColumnPolymer based mixed mode anion exchange0.5 mm
CH-953287TurboFlow Cyclone MCX ColumnPolymer based phase mixed cation exchange0.5 mm
CH-953288TurboFlow Cyclone ColumnPolymer based0.5 mm
CH-953289TurboFlow Cyclone-P ColumnPolymer based0.5 mm
CH-953456TurboFlow Cyclone MCX-2 ColumnPolymer based phase mixed cation exchange1 mm
CH-953457TurboFlow Cyclone MCX-2 ColumnPolymer based phase mixed cation exchange0.5 mm
Catalog number CH-953285
Price (USD)
835.08
Each
Description:
TurboFlow Cyclone C2 Column
Column Phase:
Silica based reversed phase
Diameter (Metric) Outer:
0.5 mm
Price (USD)
835.08
Each
Use Thermo Scientific™ TurboFlow™ Columns for automated online matrix cleanup of complex matrices analyzed on the Thermo Scientific™ Aria™ and Transcend™ TLX. Placed before the analytical column, TurboFlow sample cleanup columns enable direct injection of complex matrices such as food, plasma, urine, and other biological matrices into the mass spectrometry (MS) system without prior sample pretreatment. The patented TurboFlow column design retains compounds of interest for subsequent analysis, while complex matrices quickly and completely elute to waste.
Improve productivity
TurboFlow columns automate the many tasks traditionally performed manually, thereby eliminating the many error-prone manual steps performed for matrix cleanup that cause variability in data. As a result, this automated sample cleanup technique can improve productivity and certainty in test results.
Minimize ion supression, produce clean sample extracts:
  • Minimize sample preparation – simplifies complex sample preparation protocols
  • Reduce ion suppression – achieves better data quality
  • Simplify method development – uses the same method for different matrices
  • Save time – injects samples directly into LC-MS system
Specifications
Column PhaseSilica based reversed phase
DescriptionTurboFlow Cyclone C2 Column
Diameter (Metric) Inner0.5 mm
Diameter (Metric) Outer0.5 mm
For Use With (Application)Multi-Functional Pharmaceuticals; Extraction of Extremely Non-Polar Constituents Difficult to Elute From C8 or C18
Green FeaturesFSC
Length (Metric)50 mm
QuantityEach
For Use With (Equipment)Transcend and Aria TLX Systems
Product LineTurboFlow
SorbentC2
TypeTurboFlow XL Column
Unit SizeEach