Human IL-1 beta Matched Antibody Pair - FAQs
Human IL-1 beta Matched Antibody Pair - FAQs
View additional product information for Human IL-1 beta Matched Antibody Pair - FAQs (CHC1213)
1 product FAQs found
Do IL-1 beta Human Matched Antibody Pair (Cat. No. CHC1213), Human IL-1 beta ELISA Kit (Cat. No. KAC1211), and Human IL-1 beta ELISA Kit (Cat. No. KHC0011) use the same antibodies?
Yes, all three kits use the same antibodies. For Human IL-1 beta ELISA Kit (Cat. No. KAC1211), the coating antibodies are fragmented to improve specificity and biotinylated to improve binding capacity.