Human IL-6 Matched Antibody Pair - FAQs
Human IL-6 Matched Antibody Pair - FAQs
View additional product information for Human IL-6 Matched Antibody Pair - FAQs (CHC1263)
1 product FAQs found
Are the same antibodies used in the following IL-6 Human ELISA Kits: - Human IL-6 ELISA Kit (Cat. No. KHC0061) - Human IL-6 ELISA Kit (Cat. No. KAC1261) - Human IL-6 Matched Antibody Pair (Cat. No. CHC1263)
Yes, all three 3 kits use the same antibodies.