PT100 External Probe
Thermo Scientific™
PT100 External Probe
For Use With: Cimarec and Supernuova Hotplates and Hotplate Stirrers
Have Questions?
Catalog NumberType
CIC0001447Probe
Catalog number CIC0001447
Price (USD)
93.92
Online exclusive
106.00
Save 12.08
Each
Type:
Probe
Price (USD)
93.92
Online exclusive
106.00
Save 12.08
Each
For Use With: Cimarec and Supernuova Hotplates and Hotplate Stirrers
  • Composed of AISI 316Ti, a titanium-stabilized version of the 316 stainless steel
  • AISI 316Ti is a type of austenitic chromium-nickel stainless steel specifically designed to resist intergranular corrosion at high temperatures
Specifications
DescriptionPT100 External probe, 0-400°C, Stainless steel, for all sizes of Cimarec+ and SuperNuova+ Hotplates and Hotplate Stirrers
DisplayDigital
For Use With (Equipment)Cimarec and Supernuova Hotplates and Hotplate Stirrers
Temperature (Metric)0°C to 400°C
MaterialStainless Steel
TypeProbe
Unit SizeEach