TRACE™ 1610 Permanent Gas Analyzers (PGA)
TRACE™ 1610 Permanent Gas Analyzers (PGA)
TRACE™ 1610 Permanent Gas Analyzers (PGA)
Thermo Scientific™
TRACE™ 1610 Permanent Gas Analyzers (PGA)
Discover the Thermo Scientific™ TRACE™ 1610 Permanent Gas Analyzers available in dedicated configurations to cover different needs for industrial gas streams analysis.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFunctionVoltage
CL2GAS160001Permanent Gas with Hydrogen in Chlorine Gas120 V
PGA160010011Permanent Gas120 V
PGA160010011-230Permanent Gas230 V
PGACOCO2161Permanent Gas with Low Level CO and CO2120 V
PGACOCO2161-230Permanent Gas with Low Level CO and CO2230 V
H2PGA1602011Permanent Gas with Hydrogen120 V
H2PGA1602011-230Permanent Gas with Hydrogen230 V
H2PGACOCO216Permanent Gas with Low Level CO, CO2, Hydrogen120 V
H2PGACOCO216-230Permanent Gas with Low Level CO, CO2, Hydrogen230 V
H2PGAINRT1612Permanent Gas with Hydrogen in Acid Gas120 V
H2PGAINRT1612-230Permanent Gas with Hydrogen in Acid Gas230 V
CL2GAS160001-230Permanent Gas with Hydrogen in Chlorine Gas230 V
Catalog number CL2GAS160001
Price (USD)
-
Function:
Permanent Gas with Hydrogen in Chlorine Gas
Voltage:
120 V
Discover our wide variety of Thermo Scientific™ TRACE™ 1610 GC analyzers for permanent gases, including low levels of CO, CO2, and hydrogen in gas samples
Rely on the performance and robustness of the variety of TRACE 1610 gas chromatography systems for the analysis of industrial gas streams. These systems can be used in laboratories and in pilot/scale-up to at-line environments. Whether you need to analyze permanent gases, hydrogen, low levels of CO/CO2, even in acid gas samples containing H2S and other sulfur compounds, there is a system to meet your requirements.
  • Single channel system for N2, O2, CO, and CO2
  • Dual channel system for N2, O2, and low-level CO/CO2
  • Dual Channel system for N2, O2, CO, CO2, HC up to C3 and H2
  • Dual Channel system for N2, O2, CO, CO2, HC up to C3 and H2 in acid gas samples
  • Dual Channel system for N2,CO2, CO, CO2, HC up to C3 and H2 in Chlorine gas samples
  • Three channels system for H2, N2, O2 and low-level CO/CO2
Ready to install and run samples
  • Fully configured and factory tested before delivery
  • Full documentation set: start-up and troubleshooting guide, detailed flow and plumbing schematics, spare parts list, and method and factory test results
Built to maximize productivity and uptime
  • Spacious independent Auxiliary Oven with easy access to valves, rotors, and actuators
  • Optional third isothermal oven for molecular sieve columns
  • GC features iConnect injector and detector modules removing downtime for troubleshooting and maintenance
  • Heated sample bulkheads ensures complete sample transfer
  • Optimized system footprint to reduce benchspace requirement
Specifications
Dimensions (L x W x H)67 x 75 x 45 cm (26.4 x 29.5 x 17.7 in.)
FunctionPermanent Gas with Hydrogen in Chlorine Gas
IncludesGas chromatograph, two TCDs, auxiliary valve oven, two 10-port gas sampling valve, one 6-port valve, inert plumbing, columns and method
Inlet TypeGas Sampling Valve
Quantity1 Ea.
Voltage120 V
Column Capacity4 Packed
Detector TypeTCD
Product TypePermanent Gas Analyzer
Unit SizeEach