Iron Sulphite Agar (Dehydrated)
Thermo Scientific™
Iron Sulphite Agar (Dehydrated)
Detect thermophilic anaerobic organisms causing sulphide spoilage in food with this medium.
Have Questions?
카탈로그 번호수량
CM0079B500 g
카탈로그 번호 CM0079B
제품 가격(KRW)
-
수량:
500 g
Detect thermophilic anaerobic organisms causing sulphide spoilage in food with Thermo Scientific™ Oxoid™ Iron Sulphite Agar (Dehydrated). The medium is a modification of Cameron Sulphite Agar developed by the National Canners Association of America1. Iron Sulphite Agar has a reduced concentration of sodium sulphite to allow improved detection of some strains of Clostridium sporogenes.
Tryptone in the medium provides nitrogen and other nutrients necessary to support bacterial growth. The presence of sulphite reducing bacteria is indicated by the formation of black colonies. These colonies form when bacteria reduce sulphite to sulphide, which reacts with iron (III) citrate to yield a black precipitate.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
For Laboratory Use Only
사양
설명Iron Sulphate Agar
수량500 g
수율For 21.7L medium
형태Powder
제품 유형Agar
Unit SizeEach