Oxoid™ Corn Meal Mycological Agar
Thermo Scientific™
Oxoid™ Corn Meal Mycological Agar
Maintain the fungal stock culture and observe chlamydispore production by Candida albicans with this mycological medium
Have Questions?
Catalog NumberQuantity
CM0103B500 g
Catalog number CM0103B
Price (ZAR)
-
Quantity:
500 g
Maintain the fungal stock culture and observe chlamydospore production by Candida albicans with Thermo Scientific™ Oxoid™ Corn Meal Mycological Agar (Dehydrated). Microscopic examination of Candida albicans on this medium shows the characteristic chlamydospore production which is an accepted criterion for the identification of this species. Corn Meal Mycological Agar, being a nutritionally impoverished medium, can be employed for the maintenance of stock cultures of fungi, especially the black pigmented varieties.
Use Corn Meal Mycological Agar for chlamydospore production by Candida albicans for the maintenance of fungal stock cultures.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
For Laboratory Use Only
Specifications
DescriptionCorn Meal Agar
Quantity500 g
YieldFor 29.4L medium
FormPowder
Product TypeAgar
Unit SizeEach