MacConkey Agar No. 3 (Dehydrated)
Actual product may vary
Thermo Scientific™
MacConkey Agar No. 3 (Dehydrated)
Detect and enumerate coliform organisms, including E. coli, while isolating and detecting Salmonella and Shigella spp. with this selective medium.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호수량
CM0115R2.5kg
CM0115K25 kg
CM0115T5 kg
CM0115B500 g
카탈로그 번호 CM0115R
제품 가격(KRW)
-
수량:
2.5kg
Detect and enumerate coliform organisms, including Escherichia coli, while isolating and detecting Salmonella and Shigella spp. in both clinical and food specimens using Thermo Scientific™ Oxoid™ MacConkey Agar No.3. A modification of the original MacConkey medium by the addition of selected bile salts, the medium gives improved differentiation of coliforms and non-lactose fermenting organisms while completely inhibiting Gram-positive cocci.
Use MacConkey Agar No. 3 for detection and enumeration of coliform organisms while isolating and detecting Salmonella and Shigella spp. in pathological and food specimens.
  • Selective and differential: due to the inclusion of a specially prepared fraction of bile salts in addition to crystal violet, the medium gives improved differentiation between coliforms and non-lactose fermenting organisms while Gram-positive cocci are completely inhibited.
  • Easy-to-read: Escherichia coli (red colonies with bile precipitation) is easy to differentiate from Shigella spp. (straw coloured colonies) and Salmonella (Straw coloured colonies).
  • Recommended by the American Public Health Association 1
MacConkey Agar No.3 formulation corresponds with that recommended by the American Public Health Association1 for the direct plating of water samples for coliform bacilli, for the examination of food samples for food poisoning organisms2 and for the isolation of Salmonella and Shigella species in cheese3.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand
For Laboratory Use Only
1. American Public Health Association (1998) Standard Methods for the Examination of Water and Wastewater. 20th Edn. APHA Inc. Washington DC.
2. American Public Health Association (1976) Compendium of methods for the Microbiological Examination of Foods. APHA Inc. Washington DC.
3. American Public Health Association (1978) Standard Methods for the Examination of Dairy Products. 14th Edn. APHA Inc. Washington DC
사양
설명MacConkey Agar No. 3 (U.S. formulation)
수량2.5kg
수율For 9.7 L mediμm
형태Powder
제품 유형MacConkey Agar
Unit SizeEach