Brucella Medium Base (Dehydrated)
Thermo Scientific™
Brucella Medium Base (Dehydrated)
Isolate and cultivate Brucella with this medium when used with Brucella Selective Supplement (SR0083 or SR0209).
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
CM0169K25 kg
CM0169B500 g
Catalog number CM0169K
Price (USD)
-
Quantity:
25 kg
Isolate and cultivate Brucella using Thermo Scientific™ Oxoid™ Brucella Medium Base (Dehydrated) when used with Brucella Selective Supplement (SR0083 or SR0209). Brucella medium Base may be used to prepare the serum-dextrose-antibiotic medium as described by Jones and Brinley Morgan1 for the cultivation and isolation of Brucella, including fastidious types.
Isolate and cultivate Brucella using Brucella Medium Base when used with Brucella Selective Supplement (SR0083 or SR0209).
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
For Laboratory Use Only
Specifications
Quantity25 kg
FormPowder
Product TypeAgar
Unit SizeEach