Pseudomonas Agar Base (Dehydrated)
Thermo Scientific™
Pseudomonas Agar Base (Dehydrated)
Selectively isolate Pseudomonasspecies from a range of samples with this medium.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
CM0559B500 g
CM0559R2.5 kg
CM0559T5 kg
Catalog number CM0559B
Price (MXN)
-
Quantity:
500 g
Selectively isolate Pseudomonas species from a range of samples with Thermo Scientific™ Oxoid™ Pseudomonas Agar Base (Dehydrated). The medium is modification of King's A Medium1 and contains magnesium chloride and potassium sulphate for improved pigment production and recovery of Pseudomonas aeruginosa while strongly suppressing Klebsiella, Proteus and Providencia spp.
Pseudomonas Agar Base is designed so that by the addition of the appropriate supplement (SR0102 or SR0103) the medium becomes selective for Pseudomonas aeruginosa or Pseudomonas spp. generally.
  • Selective: Pseudomonas aeruginosa forms fluorescent green colonies while the growth of Klebsiella, Proteus, and Providencia is strongly suppressed
  • Enhanced pigment production aids identification
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand
Specifications
DescriptionPseudomonas Agar Base
Quantity500 g
YieldFor 10.3L medium
FormPowder
Product TypeAgar
Unit SizeEach