Dichloran Rose-Bengal Chloramphenicol (DRBC) Agar Base (Dehydrated)
Dichloran Rose-Bengal Chloramphenicol (DRBC) Agar Base (Dehydrated)
Dichloran Rose-Bengal Chloramphenicol (DRBC) Agar Base (Dehydrated)
Thermo Scientific™
Dichloran Rose-Bengal Chloramphenicol (DRBC) Agar Base (Dehydrated)
Isolate and enumerate yeasts and molds associated with food spoilage with this selective medium.
Have Questions?
Catalog NumberQuantity
CM0727B500 g
Catalog number CM0727B
Price (USD)
-
Quantity:
500 g
Isolate and enumerate yeasts and molds associated with food spoilage with Thermo Scientific™ Oxoid™ Dichloran Rose-Bengal Chloramphenicol (DRBC) Agar Base (Dehydrated). DRBC Agar Base is specially designed to supress the growth of spreading mold and bacteria while supporting the growth of species that cannot be isolated on Rose Bengal Chloramphenicol Agar or Acidified Potato Dextrose Agar1.
Dichloran Rose-Bengal Chloramphenicol (DRBC) Agar Base is based on the formulation described by King et al.1,2, and is recommended as a selective medium for the isolation and enumeration of yeasts and molds that are of significance in food spoilage.
  • Easy-colony-counting: Rose-Bengal in the medium controls the size and height of mold colonies preventing overgrowth and making colony counting easier
  • Selective: appropriate for isolation of yeasts and molds from foods whose associated flora consists mainly of Gram-negative rod-shaped bacteria
DRBC Agar Base is a modification of Rose-Bengal Chloramphenicol Medium3 and differs as follows: pH is lowered to 5.6, the Rose-Bengal content is reduced by 50% and Dichloran is added.
The cumulative effect of these modifications is to further inhibit bacterial growth, inhibit spreading molds such as Rhizopus and Mucor and make the medium capable of supporting the growth of those species that cannot be isolated on Rose-Bengal Chloramphenicol Agar or acidified Potato Dextrose Agar1. The inhibition of spreading molds and the general restriction of colony size results in improved enumeration and detection of mycotoxigenic molds and other species of significance in food spoilage6.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
For Laboratory Use Only
General References:
  1. King D. A. Jr., Hocking A. D. and Pitt J. I. (1979) J. Appl. & Environ. Microbiol. 37. 959-964.
  2. Pitt J. I. (1984) Personal Communication.
  3. Jarvis B. (1973) J. Appl. Bact. 36. 723-727.
  4. Thomson G. F. (1984) Food Microbiol. 1. 223-227
Specifications
Certifications/ComplianceIndustrial Reference: NMKL standards.
DescriptionDRBC Agar Base
Quantity500 g
YieldFor 15.9L medium
FormPowder
Product TypeAgar
Unit SizeEach