GBS Agar Base (Islam) (Dehydrated)
Thermo Scientific™
GBS Agar Base (Islam) (Dehydrated)
Isolate and detect Group B streptococci (GBS) in clinical specimens with this medium.
Have Questions?
Catalog NumberQuantity
CM0755B500 g
Catalog number CM0755B
Price (USD)
-
Quantity:
500 g
Isolate and detect Group B streptococci (GBS) in clinical specimens with Thermo Scientific™ Oxoid™ GBS Agar Base (Islam) (Dehydrated). Based on the formulation described by Islam1 the medium is designed to exploit the ability of most group B streptococci (GBS) to produce orange/red pigmented colonies when incubated under anaerobic conditions.
Group B streptococci are a recognized cause of serious neonatal infection acquired from the infected mother. A review2 of national data over an 8-year period by the Public Health Laboratory Service showed that group B streptococci accounted for 29.5% of all reports of neonatal bacterial meningitis with organisms being isolated from CSF and blood. Group B streptococci may also be isolated from adults infected in a variety of sites.
Use GBS Agar Base for isolation and detection Group B streptococci in clinical specimens.
  • Easy-to-read: clear colonies of group B streptococci are 0.5-1mm in diameter, round, entire and pigmented orange/red after 24-48 hours anaerobic incubation.
  • Selective: selective for Group B streptococci.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand
For Laboratory Use Only
General References:
  1. Islam A. K. M. S. (1977) Lancet i 256-257 (letter).
  2. PHLS Communicable Disease Report (1954) CDR 84/38, 3-6.
Specifications
DescriptionGBS Agar Base
Quantity500 g
YieldFor 11.1L medium
FormPowder
Product TypeAgar
Unit SizeEach