Aeromonas Medium Base (Ryan) (Dehydrated)
Thermo Scientific™
Aeromonas Medium Base (Ryan) (Dehydrated)
Isolate Aeromonas hydrophila from clinical and environmental specimens with this selective, diagnostic medium.
Have Questions?
Catalog NumberQuantity
CM0833B500 g
Catalog number CM0833B
Price (KRW)
-
Quantity:
500 g
Isolate Aeromonas hydrophila from clinical and environmental specimens with selective, diagnostic Thermo Scientific™ Oxoid™ Aeromonas Medium Base (Ryan) (Dehydrated) when used with Ampicillin Selective Supplement (SR0136).
Ryan1 modified the formulation of XLD Medium so that it would support the growth of Aeromonas spp and Plesiomonas spp as well as the usual Enterobacteriaceae. It could therefore be used as a universal medium in the investigation of enteric disease. To improve its performance in the isolation of aeromonads, the addition of ampicillin at 5mg/L is recommended.
Aeromonas spp occur widely in soil and water, where they cause diseases in fish and amphibians. They also occur in untreated and chlorinated drinking water, raw foods and raw milk.2,3It is considered that the major cause of gastrointestinal infections by Aeromonas spp3,4 is from ingesting infected water.4,5
Aeromonas Medium Base has been developed to improve the enumeration and isolation of Aeromonas spp from clinical and environmental specimens.
  • Specified by the MAFF/DHS Steering Group on the Microbiological Safety of Food for detection and enumeration of Aeromonas hydrophilain clinical specimens7.
  • Superior performance over other formulae for detection of Aeromonas in tap water, bottled water and foods including meat, poultry, fish and seafoods8,9,10
To improve the performance of the medium in the isolation of aeromonads, the addition of ampicillin at 5mg/L is recommended. The effectiveness of ampicillin as a selective agent for Aeromonas spp has been reported by several workers 11,12,13,14,15. The value of Aeromonas Medium Base (Ryan) is that the recommended level of ampicillin is well below that which can cause inhibition of some strains of aeromonad16.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand
For Laboratory Use Only
General References:
  1. Ryan N. (1985) Personal communication
  2. Steering Group on the Microbiological Safety of Food (SGMSF). Methods for use in Microbiological Surveillance (1994) MAFF. Ergon House London SW1P 3TR.
  3. Buchanan R.L. and Palumb S.A. (1985) J. Food Safety 7. 1
  4. Burke V., Robinson J., Gracey M., Peterson D. and Partridge K. (1984) Appl. Environ. Microbiol. 48. 361-366.
  5. George W.L. (1987) Clin. Microbiol. Newsletter 9. 121-122.
  6. Holmberg S.D., Schell W.L., Fanning G.R., Wachsmith L.K., Hickman-Brenner F.W., Blake
  7. Steering Group on the Microbiological Safety of Food (SGMSF). Methods for use in Microbiological Surveillance (1994) MAFF. Ergon House London SW1P 3TR
  8. Holmes P. and Sartory D.P. (1993) Letters in Applied Microbiol. 17. 58-60.
  9. C. Pin M.L., Marin M.L., Garcia J et al (1994) Letters in Appl. Microbiol. 18. 190-192.
  10. Warburton D.W., McCormick J.K. and Bowen B. (1994) Can. J. Microbiol. 40. 145-148
  11. Moulsdale M.T. (1983) The Lancet i: 351.
  12. Rogol M., Sechter I., Grinberg L., Gerichter Ch. B. (1992) J. Med. Microbiol. 12. 229-231.
  13. Richardson C.J.L., Robinson J.O., Wagener L.B. and Burke V. (1982) J. Antimicrob. Chemother. 9. 267-274.
  14. Atkinson M. (1986) Culture Vol. 7, No.2.
  15. Mishra S., Nair G.B., Bhadra R.K., Sikder S.N. and Pal. S.C. (1987) J. Clin. Microbiol. 25. 2040-2043.
  16. Rahim Z., Sanyal S.C., Aziz K.M.S., Huq M.I. and Chowbury A.A. (1984) Appl. Environ. Microbiol. 48. 865-867
Specifications
DescriptionAeromonas Medium Base (Ryan)
Quantity500 g
YieldFor 8.5L medium
FormPowder
Product TypeDehydrated Culture Media
Unit SizeEach