VRE Broth Base (Dehydrated)
VRE Broth Base (Dehydrated)
Thermo Scientific™
VRE Broth Base (Dehydrated)
Isolate Vancomycin-Resistant Enterococci (VRE) and High-Level Aminoglycoside Resistant Enterococci (HLARE) from clinical samples with this medium.
Have Questions?
Número de catálogoCantidad
CM0984B500 g
Número de catálogo CM0984B
Precio (MXN)
-
Cantidad:
500 g
Isolate Vancomycin-Resistant Enterococci (VRE) and High-Level Aminoglycoside Resistant Enterococci (HLARE) from clinical samples with Thermo Scientific™ Oxoid™ VRE Broth Base (Dehydrated). The proliferation of enterococci resistant to many commonly used antimicrobials is on the increase1. The recent emergence of VRE is of great concern as enterococci can cause bacteremia, endocarditis, and urinary tract infections.
The use of VRE Agar Base complies with recommendations from the Center for Disease Control and Prevention (CDC) to detect VRE infection in its early stages2.
The proliferation of enterococci, resistant to many commonly used antimicrobials is on the increase1. The recent emergence of VRE is of great concern as enterococci can cause bacteremia, endocarditis, and urinary tract infections.
The use of VRE Agar Base complies with recommendations from the Center for Disease Control and Prevention (CDC) to detect VRE infection in its early stages2.
VRE Agar Base contains an indicator system to detect the growth of aesculin-hydrolysing organisms. Enterococci produce black zones around the colonies from the formation of black iron phenolic compounds derived from aesculin-hydrolysis products and ferrous iron.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Solo para uso en laboratorio
General References:
  1. King, W. K. (1996) Bug Bytes Vol. 2 No. 19.
  2. CDC Preventing the spread of vancomycin resistance: a report from the Hospital Infection Control Practices Advisory Committee (1994). Fed Regist. May 17.
Especificaciones
DescripciónVRE Broth Base
Cantidad500 g
ProducciónFor 13.5L medium
FormularioPolvo
Tipo de productoDehydrated Culture Media
Unit SizeEach