<i>Brilliance</i>&trade; UTI Clarity&trade; Agar (Dehydrated)
Thermo Scientific™

Brilliance™ UTI Clarity™ Agar (Dehydrated)

Isolate, enumerate, and presumptively identify urinary pathogens with this chromgenic medium.
Have Questions?
Alterar visualizaçãobuttonViewtableView
Número do catálogoFormQuantity
CM1106TPowder5 kg
CM1106BPowder500 g
Número do catálogo CM1106T
Preço (BRL)
-
Form:
Powder
Quantity:
5 kg
Easily isolate, enumerate, and presumptively identify common organisms associated with urinary tract infections using Thermo Scientific™ Brilliance™ UTI Clarity™ Agar. Organisms are differentiated through the utilization of two chromogenic substrates and/or tryptophan.

The Brilliance UTI Clarity Agar formulation contains two chromogenic substrates which are cleaved by the ß-galactosidase and ß-glucosidase enzymes produced by E. coli, Enterococcus species, and coliforms. These specific enzyme reactions cleave the chromogens, giving a range of diagnostic colors. This medium targets the same enzymes as other chromogenic agar: Part No. CM1050B, Chromogenic UTI medium. However, to do so, new and more specific chromogens have been incorporated. The new chromogens provide improved reactions, enabling even easier differentiation of organism groups:

  • Easy-to-read: the ß-galactosidase activity of E. coliand S. saprophyticus results in pink/pale pink colonies, and the ß-glucosidase activity of enterococci produces blue/turquoise colonies; the activity of both enzymes on coliforms gives dark blue/purple colonies. Tryptophan deaminase activity produces a brown halo around colonies of Proteus, Morganella, and Providencia species. Most other organisms exhibit their natural pigmentation
  • Reduced confirmatory tests: the specificity of Brilliance UTI Clarity Agar minimises confirmatory testing
  • Rapid: same day results for E. coli confirmation being achieved using rapid indole test (DMAC) (Part No. MB1448A)

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

For In Vitro Diagnostic Use.  
Especificações
Quantity5 kg
Shelf Life5 Years
FormPowder
Product TypeAgar
Unit SizeEach
Conteúdo e armazenamento
10°C to 30°C