24 Listeria Enrichment Broth (24 LEB) (Dehydrated)
24 Listeria Enrichment Broth (24 LEB) (Dehydrated)
24 Listeria Enrichment Broth (24 LEB) (Dehydrated)
Thermo Scientific™
24 Listeria Enrichment Broth (24 LEB) (Dehydrated)
Selectively enrich Listeria monocytogenes and other Listeria spp. from food and environmental samples.
Have Questions?
Catalog NumberQuantity
CM1107B500 g
Catalog number CM1107B
Price (MXN)
-
Quantity:
500 g
Selectively enrich Listeria monocytogenes and other Listeria spp. from food and environmental samples with Thermo Scientific™ Oxoid™ 24 Listeria Enrichment Broth (24 LEB) (Dehydrated). The medium provides optimal growing conditions for all Listeria species for PCR applications. It is specifically designed for use with PCR systems, such as Thermo Scientific™ SureTect™ molecular assays for Listeria.
Listeria monocytogenes is the most common pathogenic Listeria spp. and has been shown to be pathogenic to both humans and animals. 24 Listeria Enrichment Broth (24 LEB) is designed to identify Listeria spp. based on their utilization of a chomogenic substrate.
  • Easy-to-read: chromogenic and phospholipase enzyme reactions easily differentiate Listeria monocytogenes from other Listeria spp.
  • Adheres to ISO 11290-1:1997 standards.
  • Superior to PALCAM and Oxford medium for the isolation of Listeria monocytogenes1.
The presence of the chromogenic substrate for the detection of  glucosidase (an enzyme common to all Listeria spp. and to a few strains of Enterococci and Bacilli) results in chromogenic activity.
Further differentiation of Listeria monocytogenes and other Listeria spp. is facilitated by the addition of phosphatidylinositol or lethicin substrate for the detection of phospholipase enzyme.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific™ brand.
Specifications
Description24 Listeria Enrichment Broth (24 LEB) (Dehydrated)
Quantity500 g
Shelf Life5 Years
FormPowder
Product TypeDehydrated Culture Media
SpeciesListeria
Unit SizeEach
Contents & Storage
10°C to 30°C