Search
Search

Cultivate streptococci, Neisseria and other fastidious organisms with this highly nutritious medium.
| Número do catálogo | Quantity |
|---|---|
| CM1135T | 5 kg |
| CM1135B | 500 g |
| CM1135K | 25 kg |
Cultivate streptococci, Neisseria and other fastidious organisms with Thermo Scientific™ Oxoid™ Brain Infusion Broth (Dehydrated). The medium is highly nutritious and well buffered to support the growth of wide variety of organisms. It is recommended for blood culture work for the isolation and cultivation of pathogenic fungi.
Cultivate a wide variety of organisms with Brain Infusion Broth:
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.