Oxoid™ 24 LEB Complete
Thermo Scientific™
Oxoid™ 24 LEB Complete
24 LEB complete is used for the enrichment of Listeria sp and is pre-supplemented with 24 LEB Selective Supplement (does not require the addition of SR0243E)
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCantidad
CM1154B1 unidad
CM1154G10 kg
Número de catálogo CM1154B
Precio (MXN)
-
Cantidad:
1 unidad
Selectively enrichment of Listeria monocytogenes and other Listeria spp. from food and environmental samples with Thermo Scientific™ 24 Listeria Enrichment Broth (24 LEB) (Dehydrated). The medium provides optimal growing conditions for all Listeria species for PCR applications. It is specifically designed for use with PCR systems, such as Thermo Scientific™ SureTect™ molecular assays for Listeria.
Listeria monocytogenes is the most common pathogenic Listeria spp. and has been shown to be pathogenic to both humans and animals. 24 Listeria Enrichment Broth (24 LEB) is designed to identify Listeria spp. based on their utilization of a chomogenic substrate. Easy-to-read: chromogenic and phospholipase enzyme reactions easily differentiate Listeria monocytogenes from other Listeria spp.Adheres to ISO 11290-1:1997 standards.The presence of the chromogenic substrate for the detection of glucosidase (an enzyme common to all Listeria spp. and to a few strains of Enterococci and Bacilli) results in chromogenic activity. Further differentiation of Listeria monocytogenes and other Listeria spp. is facilitated by the addition of phosphatidylinositol or lethicin substrate for the detection of phospholipase enzyme.
Especificaciones
Certificaciones/conformidadAdheres to ISO 11290-1:1997 standards.
País de origenUK
Para utilizar con (aplicación)Selective Enrichment of Listeria sp From Food and Environmental Samples
Tipo de mediosDehydrated Culture Media
Tipo de empaquetadoTub
Cantidad1 unidad
Duración de almacenamiento2-5 years
Tipo de productoDehydrated Culture Media
EspecieListeria
Unit SizeEach
Contenido y almacenamiento
Room Temperature