COD (Chemical Oxygen Demand) Calibration Standards for AQUAfast Colorimeters
Thermo Scientific™

COD (Chemical Oxygen Demand) Calibration Standards for AQUAfast Colorimeters

Use these calibration standards to check your colorimeter instrument operations before measuring COD chemistries, for example in environmental and wastewater monitoring.
Have Questions?
Catalog NumberConcentration
CODS1010000 ppm
Catalog number CODS10
Price (USD)
74.88
Online exclusive
83.20
Save 8.32
Each
Concentration:
10000 ppm
Request bulk or custom format
Price (USD)
74.88
Online exclusive
83.20
Save 8.32
Each
Specifications
For Use With (Equipment)AQUAfast Colorimeter
Quantity475 mL
Certifications/ComplianceEPA
Concentration10000 ppm
Industry TypeEnvironmental Testing, Wastewater Monitoring
View more

Use Thermo Scientific™ COD (Chemical Oxygen Demand) Calibration Standards with Orion™ AQUAfast Colorimeters such as AQ4000 Advanced Colorimeter and Orion AQUAfast II AQ2040 Colorimeter. Orion colorimeters, which measure COD chemistries, are ideal for environmental and wastewater monitoring. Use these calibration standards to check operations prior to using the instruments.

  • For use with: Orion AQUAfast AQ4000, Orion AQUAfast AQ2040 colorimeters
  • Two Calibration Standards are available: 1000 ppm (CODS01) and 10000 ppm (CODS10), both in 475 mL bottles
  • Ideal for environmental and wastewater monitoring