X-ray Inspection and Checkweigher Combination Unit
X-ray Inspection and Checkweigher Combination Unit
X-ray Inspection and Checkweigher Combination Unit
X-ray Inspection and Checkweigher Combination Unit
X-ray Inspection and Checkweigher Combination Unit
Thermo Scientific™
X-ray Inspection and Checkweigher Combination Unit
Our x-ray inspection and checkweigher combination can be installed within production and packaging lines for food and beverage, pharmaceutical, or personal care product manufacturers. These combination systems provide many benefits versus purchasing separate systems, such as providing space savings and budget savings to manufacturers as well as a single point for maintenance and quality control.
Have Questions?
Catalog NumberProduct Type
COMBOXRAYX-ray Inspection /Checkweigher
Catalog number COMBOXRAY
Price (KRW)
-
Product Type:
X-ray Inspection /Checkweigher
Our Thermo Scientific x-ray and checkweigher combination system includes the Thermo Scientific™ NextGuard™ C330 X-ray inspection and the Thermo Scientific™ Global Versa checkweigher systems with integrated conveyor belts and dual rejection systems.
The NextGuard C330 is ideal for finding dense or sharp objects in a wide variety of products as well as detecting errors like over/under fill or missing, misplaced, cracked, or incorrectly formed products. The Versa checkweigher has proprietary software with advanced automatic-zero algorithms to ensure accuracy standards are maintained over time, regardless of environment, and is engineered for ease of maintenance.
There are many benefits to combining x-ray inspection and a checkweigher into one system, providing maximum product safety and the protection of your brand.
  • Single point of operation for personnel
    • Many companies have quality control procedures where critical control points within their production process need to be monitored. Having the x-ray inspection combined with the checkweigher, enables both systems to easily be monitored by the employee on the production floor and not have to move a great distance between the two systems.
    • Reduces safety risk to the operator as the reject systems of both the x-ray system and checkweigher system are located next to each other.
  • Single point for maintenance
    • Set-up and calibration of the x-ray and checkweigher systems can be done together at the same time.
  • Space saving
    • Production facilities are often very crowded and constrained by lack of extra space. Even a space savings of 200 cm is valuable within a constricted manufacturing plant. This combination system can save from 200 cm to 1,000 cm in length of valuable packaging line space.
  • Budget saving
    • Combination units save on the purchase of hardware. Since the conveyor belt of the NextGuard x-ray is also used as the infeed for the Versa checkweigher this eliminates the need for an extra conveyor belt which can be a cost savings of several thousand dollars compared to buying two separate systems.
  • Single point for quality control
    • Rejection of out-of-range weight and contaminated products are side by side for easy removal.
    • Each system has its own HMI control screen, and both are visible from a single point allowing simultaneous review of real time data from both systems.
    • X-ray belt can help pull pitch of products, regulating product presentation going onto the checkweigher.
Specifications
Product TypeX-ray Inspection /Checkweigher
Unit SizeEach