Product Image
Invitrogen™
ChargeSwitch™ Lysis Buffer L13
The ChargeSwitch® Forensic DNA Purification Kit is a magnetic bead method designed to accommodate all forensic crime scene sample typesRead more
Have Questions?
Catalog NumberQuantity
CS11202800 mL
Catalog number CS11202
Price (USD)
421.00
Each
Quantity:
800 mL
Price (USD)
421.00
Each
The ChargeSwitch® Forensic DNA Purification Kit is a magnetic bead method designed to accommodate all forensic crime scene sample types for STR profiling. The kit is scalable for single-tube use or for automated extraction.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
BufferLysis Buffer
For Use With (Application)DNA Extraction
Product LineChargeSwitch
Product TypeLysis Buffer L13
Quantity800 mL
Shipping ConditionRoom Temperature
FormatKit
Unit SizeEach
Contents & Storage
All components of the ChargeSwitch™ Forensic DNA Purification Kits are shipped at room temperature. Upon receipt, store the Proteinase K at 4°C. Store all other components at room temperature. All components are guaranteed stable for 6 months, if stored properly.
Frequently asked questions (FAQs)
I used water to elute my plasmid instead of the Elution Buffer with my ChargeSwitch kit, but saw very low plasmid yield. Why is this?
We strongly recommend using the Elution Buffer provided in the kit, and do not recommend elution with water. If you need to elute in any other buffer, be sure to use a buffer of pH 8.5-9.0 for efficient DNA elution.
What is the difference between the ChargeSwitch Pro kit and the Charge-Switch Pro filter kit?
The filter kit is based upon a dual nested column design, which facilitates the fastest and simplest protocol for obtaining high quality plasmid (less than 10 EU/µg DNA). Modified wash buffer 1 removes more endotoxin.
What is the typical yield when using the ChargeSwitch Pro Filter Plasmid Mini or Midi Kits?
The typical yield is 25 µg from a 5 mL culture for the ChargeSwitch Pro mini kit, 200 µg from a 25 mL culture for the midi kit, and 800 µg from a 100 mL culture from the maxi kit. The plasmid size range for the kits is typically 3-9 kb.
How does ChargeSwitch technology work?
Based on a patented switchable chemistry, ChargeSwitch purification utilizes the charge on a solid surface for nucleic acid capture. At lower pH, the surface chemistry holds a positive charge and attracts nucleic acid. At higher pH, the surface chemistry is neutral and any bound nucleic acid is released.