Product Image
Invitrogen™

ChargeSwitch™-Pro PCR Clean-Up Kit

The ChargeSwitch™-Pro PCR Clean-up Kit purifies high-quality DNA fragments without the use of ethanol or harsh salts known to interfereRead more
Catalog NumberNo. of Reactions
CS3205050 Preps
CS32250250 Preps
Catalog number CS32050
Price (JPY)
13,800
Online offer
Ends: 25-Sep-2026
23,100
Save 9,300 (40%)
Each
No. of Reactions:
50 Preps

The ChargeSwitch™-Pro PCR Clean-up Kit purifies high-quality DNA fragments without the use of ethanol or harsh salts known to interfere with enzymatic reactions.

The ChargeSwitch™-Pro Kit provides:
• 100% ethanol-free, guanidinium-free, organic solvent-free protocols
• Improved downstream performance
• Complete kit for fast, hassle-free setup
• 20% fewer handling steps than most PCR clean-up protocols
• Purification of up to 500 μL of PCR product in one column


How It Works
ChargeSwitch™ technology employs a completely novel mechanism that replaces harsh reagents such as chaotropic salts, ethanol, or organic solvents and isopropanol precipitation with a simple ion-exchange procedure. In this method, DNA binds to a surface ligand that is positively charged at low pH, and is eluted when the pH is raised to 8.5 (Figure 1). The ChargeSwitch™-Pro protocol retains the ease and familiarity of common silica spin-column protocols with binding, washing, and elution steps (Figure 2).


Better Downstream Results
Since ChargeSwitch™ technology does not require the use of ethanol, chaotropic salts, or organic solvents for nucleic acid purification, the resulting DNA is free of enzymatic inhibitors and thus shows better downstream performance (Figure 3). These compounds are known to inhibit many enzymatic reactions such as PCR, cloning, restriction enzyme digestion, and array CGH labeling. Expect to see reduced variability in your sequencing applications, with reads in excess of 800 bp with Phred scores >20 (Figure 4); clean restriction enzyme digestions (Figure 5); and removal of dNTPs and primers in excess of 98% (Figure 6).

For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormatKit
High-throughput CompatibilityNot High-throughput Compatible (Manual)
Isolation TechnologyChargeSwitch™ Chemistry
No. of Reactions50 Preps
Product LineChargeSwitch
Product TypePCR Clean-Up Kit
Quantity50 Preps
Sample TypePCR Products
Shipping ConditionRoom Temperature
TargetDNA from Enzymatic Reactions (e.g. PCR)
For Use With (Application)Next-Generation Sequencing (NGS), Cloning, Standard PCR
Unit SizeEach
Contents & Storage

The ChargeSwitch™-Pro PCR Clean-up Kit contains columns, collection tubes, recovery tubes, binding buffer, wash buffer, and elution buffer.

Store at room temperature. Guaranteed stable for 6 months when properly stored.

Frequently asked questions (FAQs)

I used water to elute my plasmid instead of the Elution Buffer with my ChargeSwitch kit, but saw very low plasmid yield. Why is this?

We strongly recommend using the Elution Buffer provided in the kit, and do not recommend elution with water. If you need to elute in any other buffer, be sure to use a buffer of pH 8.5-9.0 for efficient DNA elution.

What is the difference between the ChargeSwitch Pro kit and the Charge-Switch Pro filter kit?

The filter kit is based upon a dual nested column design, which facilitates the fastest and simplest protocol for obtaining high quality plasmid (less than 10 EU/µg DNA). Modified wash buffer 1 removes more endotoxin.

What is the typical yield when using the ChargeSwitch Pro Filter Plasmid Mini or Midi Kits?

The typical yield is 25 µg from a 5 mL culture for the ChargeSwitch Pro mini kit, 200 µg from a 25 mL culture for the midi kit, and 800 µg from a 100 mL culture from the maxi kit. The plasmid size range for the kits is typically 3-9 kb.

I'm seeing bead carryover with the ChargeSwitch PCR Clean-up Kit. Is this okay for my PCR?

ChargeSwitch coated beads are inert and will not affect PCR. We have found that volumes of as much as 10 µL of beads have no deleterious effects on regular PCR when spiked in. Excessive quantities greater than 10 µL of beads will start to inhibit PCR. Some specific applications may be affected by beads, including real-time reactions or MALDI-TOF. In these applications, repeat the final elution binding as described in the protocol.

How does ChargeSwitch technology work?

Based on a patented switchable chemistry, ChargeSwitch purification utilizes the charge on a solid surface for nucleic acid capture. At lower pH, the surface chemistry holds a positive charge and attracts nucleic acid. At higher pH, the surface chemistry is neutral and any bound nucleic acid is released.