Invitrogen™

ChargeSwitch™ Pro Filter BAC Buffer Kit

Catalog number:  CS32100

 Related applications: DNA Extraction

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

ChargeSwitch® Pro Filter BAC Buffer Kit is a convenient buffer kit enabling to separately purchase all buffers needed for the ChargeSwitch® Pro Filter Plasmid kits. The buffer set comes with three buffers (resuspension-, lysis-, and precipiatation buffers). In addition, RNase A is provided (950 µl; concentration: 20 mg/ml). This buffer kit is compatible with all ChargeSwitch® Pro Filter Plasmid kits (CS31107, CS31106, CS31104).
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Sample Type (General): Bacteria
High Throughput Compatibility: Not High Throughput-Compatible (Manual)
Downstream Application: Cloning, In Vitro Transcription, Next-Generation Sequencing, PCR, Sequencing, Transfection, Transformation
Final Product: Plasmid DNA
Isolation Technology: ChargeSwitch® Chemistry
Product Line: ChargeSwitch®
Product Size: 1 kit
Sample Type (Specific): Bacterial Culture
Starting Material (Volume): 1 ml
Yield (up to): 20 µg
Shipping Condition: Room Temperature

Contents & storage

ChargeSwitch®-Pro Resuspension Buffer: 185 ml
ChargeSwitch®-Pro Lysis Buffer: 185 ml
ChargeSwitch®-Pro Precipitation Buffer: 185 ml
ChargeSwitch®-Pro RNase: 0.95 ml

Documents

Manuals & protocols