Oxoid™ Moxifloxacin Antimicrobial Susceptibility discs
Oxoid™ Moxifloxacin Antimicrobial Susceptibility discs
Oxoid™ Moxifloxacin Antimicrobial Susceptibility discs
Thermo Scientific™
Oxoid™ Moxifloxacin Antimicrobial Susceptibility discs
Determine the susceptibility of microorganisms using manual AST methods and a full range of high-quality, easy-to-use antimicrobial susceptibility testing discs.
Have Questions?
Catalog NumberConcentration
CT1633B5 μg
Catalog number CT1633B
Price (USD)
-
Concentration:
5 μg
Manually determine the antibiotic susceptibility of microorganisms using Antimicrobial Susceptibility Testing (AST) methods in conjunction with reliable, easy-to-use Thermo Scientific™ Oxoid™ Moxifloxacin Antimicrobial Susceptibility discs.
Ease-of-use is provided by shatterproof spring-loaded cartridges, easy-to-read labeling and end-of-cartridge indicators.
disc diffusion not only categorizes resistant, intermediate and sensitive organisms through quantitative results but also provides a visual indication of the following:
  • Inoculum level
  • Presence of contamination
  • Resistant mutants
  • Beta-lactamase activity
  • Antagonism and synergy between antibiotics
Each cartridge is individually sealed with a desiccant to maintain a moisture level of <2%, ensuring long term stability.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Antibiotic CodeMXF
Antimicrobial AgentsMoxifloxacin
Certifications/ComplianceComplies with CLSI, EUCAST, SFM guidelines
Concentration5 μg
DescriptionOxoid Moxifloxacin Antimicrobial Susceptibility Discs
No. per Pack5 Cartridges/Pk.
Packaging Type50 Discs/Cartridge
Quantity50 Discs/Cartridge
Product TypeAntimicrobial Susceptibility Disc
Test TypeAntimicrobial Susceptibility
Unit SizeEach