Deionizer Cartridges
Deionizer Cartridges
Deionizer Cartridges
Deionizer Cartridges
Deionizer Cartridges
Thermo Scientific™
Deionizer Cartridges
Choose from an array of Thermo Scientific™ Deionizer Cartridges for select Thermo Scientific™ Barnstead™ water systems, based on your application.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)Resin TypeCapacityFor Use With (Application)
D0809Bantam™ Deionizer, B-Pure™Mixed Bed875 grains
D0760Bantam™ Deionizer, B-Pure™Strong Anion Exchange1680 grainsAnion Removal
D8815B-Pure™Activated Carbon5000 gal. (as CaCO3)Organic Removal
D8816B-Pure™Mixed BedUltrapure
D8818B-Pure™Ultrapure, Oxygen RemovalUltrapure, Oxygen and Organic Removal
D8825B-Pure™Mixed Bed, Ultrapure, Oxygen and Organic RemovalUltrapure and Oxygen Removal Resin
D0811B-Pure™Porous Strong Ion Exchange30 gOxygen Removal
D0813Bantam™ Deionizer, B-Pure™Activated Carbon5000 gal.Organic Removal
D0815Bantam™ Deionizer, B-Pure™Strong Cation Exchange3170 grainsCation Removal
D0832Bantam™ Deionizer, B-Pure™Mixed Bed, Activated Carbon785 grains/1000 gal.Ultrapure and Organic Removal
D0835B-Pure™Macroreticular, Activated Carbon5000 gal.Pretreatment
D0836B-Pure™Macroreticular, Activated Carbon2000 gal.Pretreatment Macropure
D50213B-Pure™Mixed Bed550 grainsUltrapure
D50214B-Pure™Porous Strong Anion Exchange18 gOxygen Removal
D50215B-Pure™Activated Carbon5000 gal.Organic Removal
D50216B-Pure™Oxygen Removal Mixed Bed275 grains/9 gOxygen and demineralization
D50217B-Pure™Mixed Bed, Activated Carbon275 grains/2000 gal.Ultrapure and Organic Removal
D63112B-Pure™Extruded Carbon5000 gal.Organic Removal
D8809B-Pure™Ultrapure, Oxygen Removal455 grains/30 gUltrapure and Organic Removal
D8811B-Pure™Ultrapure, Oxygen and Organic Removal365 grains/2000 gal./12 gUltrapure, Oxygen and Organic Removal
D0803Thermo Scientific™ Barnstead™ Bantam™ B-Pure™ or Nanopure™ Water Purification Systems as IndicatedTwo Bed Ion Exchange1760 grainsHigh capacity, two-bed ion exchange
Catalog number D0809
Price (USD)
216.00
Online exclusive
240.00
Save 24.00
Each
For Use With (Equipment):
Bantam™ Deionizer, B-Pure™
Resin Type:
Mixed Bed
Capacity:
875 grains
Price (USD)
216.00
Online exclusive
240.00
Save 24.00
Each
Choose from an array of Thermo Scientific™ Deionizer Cartridges based on your application needs. These cartridges are for use with the Thermo Scientific™ Barnstead™ Bantam™, B-Pure™ or Nanopure™ water purification systems as indicated.
  • Choice of full or half cartridges
Specifications
For Use With (Equipment)Bantam™ Deionizer, B-Pure™
TypeDeionizer Cartridge
Capacity875 grains
Resin TypeMixed Bed
Unit SizeEach