Do you offer an alternative for the discontinued 5-dodecanoylaminofluorescein (Cat. No. D109)?
Although our product 5-dodecanoylaminofluorescein (Cat. No. D109) has been discontinued, it is available from another vendor. You may order it through our affiliate Fisher Scientific linked below:
AdipoGen 5-Dodecanoylaminofluorescein (Cat. No. 50-114-7018)
AdipoGen 5-Dodecanoylaminofluorescein (Cat. No. 50-114-7018)