DQ™ Ovalbumin - Special Packaging - FAQs
DQ™ Ovalbumin - Special Packaging - FAQs
View additional product information for DQ™ Ovalbumin - Special Packaging - FAQs (D12053)
1 product FAQs found
Do you have an alternative for DQ Ovalbumin – Special Packaging (Cat. No. D12053) that can be purchased in the EU?
Unfortunately, DQ Ovalbumin – Special Packaging (Cat. No. D12053) is suspended for sale in the EU until further notice and there is no alternative for this product currently.