Thermo Scientific™

Barnstead™ RO System

Catalog number: D12671
Thermo Scientific™

Barnstead™ RO System

Catalog number: D12671
This product has been discontinued. For additional information or alternate products please contact your sales representative. The recommended replacement for this product is the Thermo Scientific™ Barnstead™ Pacific™ RO water purification system.
 
Catalog Number
D12671
Unit Size
-
Flow Rate
6 L/hr.
Price (USD)
Full specifications
Bacterial Content99%
Certifications/ComplianceCE, CSA
Chlorine Tolerance1000 ppm/hr.
Contaminant Rejection (Microbiological)99%
Contaminant Rejection (Minimum Ionic)Monovalent: 90 to 95%; Polyvalent: 95 to 99
Contaminant Rejection (Organic)99%
Contaminant Rejection (Particulate)90 to 95%
Depth (English)16.9 in.
Depth (Metric)42.9 cm
Electrical Requirements100/240 VAC 50/60 Hz
Feedwater Pressure30 to 100 psig (2 to 6.9 bar)
Frequency50/60 Hz
Height (English)19.5 in.
Height (Metric)49.5 cm
Iron Total as Fe<0.5ppm
Max. TDS as CaCO3800 ppm
Particles >0.2µM/mL<1 μM/mL
Resistivity18.2MΩ·cm
Silica<30ppm
Silt Density Index<5%
Temperature Range (English)34°F to 95°F
Temperature Range (Metric)1°C to 35°C
Turbidity<1.0 NTU
Voltage100/240 VAC
Width (English)13.5 in.
Width (Metric)34.3 cm
Feed Water MonitoringNo
Feedwater SourceTap
Flow Rate6 L/hr.
Product Water TypeUltrapure (Type 1)
TOC Monitor with UV Intensity MonitoringNo
TypeReverse Osmosis System
pHOptimum Range: 6.5 to 7.5, Operating Range: 4 to 11
Unit Size-
Catalog Number
Unit Size
-
Flow Rate
12 L/hr.
Price (USD)
Full specifications
Bacterial Content99%
Certifications/ComplianceCE, CSA
Chlorine Tolerance1000 ppm/hr.
Contaminant Rejection (Microbiological)99%
Contaminant Rejection (Minimum Ionic)Monovalent: 90 to 95%; Polyvalent: 95 to 99
Contaminant Rejection (Organic)99%
Contaminant Rejection (Particulate)90 to 95%
Depth (English)16.9 in.
Depth (Metric)42.9 cm
Electrical Requirements100/240 VAC 50/60 Hz
Feedwater Pressure30 to 100 psig (2 to 6.9 bar)
Frequency50/60 Hz
Height (English)19.5 in.
Height (Metric)49.5 cm
Iron Total as Fe<0.5ppm
Max. TDS as CaCO3800 ppm
Particles >0.2µM/mL<1 μM/mL
Resistivity18.2MΩ·cm
Silica<30ppm
Silt Density Index<5%
Temperature Range (English)34°F to 95°F
Temperature Range (Metric)1°C to 35°C
Turbidity<1.0 NTU
Voltage100/240 VAC
Width (English)13.5 in.
Width (Metric)34.3 cm
Feed Water MonitoringNo
Feedwater SourceTap
Flow Rate12 L/hr.
Product Water TypeUltrapure (Type 1)
TOC Monitor with UV Intensity MonitoringNo
TypeReverse Osmosis System
pHOptimum Range: 6.5 to 7.5, Operating Range: 4 to 11
Unit Size-
Catalog Number
Unit Size
-
Flow Rate
24 L/hr.
Price (USD)
Full specifications
Bacterial Content99%
Certifications/ComplianceCE, CSA
Chlorine Tolerance1000 ppm/hr.
Contaminant Rejection (Microbiological)99%
Contaminant Rejection (Minimum Ionic)Monovalent: 90 to 95%; Polyvalent: 95 to 99
Contaminant Rejection (Organic)99%
Contaminant Rejection (Particulate)90 to 95%
Depth (English)16.9 in.
Depth (Metric)42.9 cm
Electrical Requirements100/240 VAC 50/60 Hz
Feedwater Pressure30 to 100 psig (2 to 6.9 bar)
Frequency50/60 Hz
Height (English)19.5 in.
Height (Metric)49.5 cm
Iron Total as Fe<0.5ppm
Max. TDS as CaCO3800 ppm
Particles >0.2µM/mL<1 μM/mL
Resistivity18.2MΩ·cm
Silica<30ppm
Silt Density Index<5%
Temperature Range (English)34°F to 95°F
Temperature Range (Metric)1°C to 35°C
Turbidity<1.0 NTU
Voltage100/240 VAC
Width (English)13.5 in.
Width (Metric)34.3 cm
Feed Water MonitoringNo
Feedwater SourceTap
Flow Rate24 L/hr.
Product Water TypeUltrapure (Type 1)
TOC Monitor with UV Intensity MonitoringNo
TypeReverse Osmosis System
pHOptimum Range: 6.5 to 7.5, Operating Range: 4 to 11
Unit Size-
Showing 3 of 3
Catalog NumberSpecificationsUnit SizeFlow RatePrice (USD)
D12671Full specifications
-6 L/hr.Request A Quote
Bacterial Content99%
Certifications/ComplianceCE, CSA
Chlorine Tolerance1000 ppm/hr.
Contaminant Rejection (Microbiological)99%
Contaminant Rejection (Minimum Ionic)Monovalent: 90 to 95%; Polyvalent: 95 to 99
Contaminant Rejection (Organic)99%
Contaminant Rejection (Particulate)90 to 95%
Depth (English)16.9 in.
Depth (Metric)42.9 cm
Electrical Requirements100/240 VAC 50/60 Hz
Feedwater Pressure30 to 100 psig (2 to 6.9 bar)
Frequency50/60 Hz
Height (English)19.5 in.
Height (Metric)49.5 cm
Iron Total as Fe<0.5ppm
Max. TDS as CaCO3800 ppm
Particles >0.2µM/mL<1 μM/mL
Resistivity18.2MΩ·cm
Silica<30ppm
Silt Density Index<5%
Temperature Range (English)34°F to 95°F
Temperature Range (Metric)1°C to 35°C
Turbidity<1.0 NTU
Voltage100/240 VAC
Width (English)13.5 in.
Width (Metric)34.3 cm
Feed Water MonitoringNo
Feedwater SourceTap
Flow Rate6 L/hr.
Product Water TypeUltrapure (Type 1)
TOC Monitor with UV Intensity MonitoringNo
TypeReverse Osmosis System
pHOptimum Range: 6.5 to 7.5, Operating Range: 4 to 11
Unit Size-
D12651Full specifications
-12 L/hr.Request A Quote
Bacterial Content99%
Certifications/ComplianceCE, CSA
Chlorine Tolerance1000 ppm/hr.
Contaminant Rejection (Microbiological)99%
Contaminant Rejection (Minimum Ionic)Monovalent: 90 to 95%; Polyvalent: 95 to 99
Contaminant Rejection (Organic)99%
Contaminant Rejection (Particulate)90 to 95%
Depth (English)16.9 in.
Depth (Metric)42.9 cm
Electrical Requirements100/240 VAC 50/60 Hz
Feedwater Pressure30 to 100 psig (2 to 6.9 bar)
Frequency50/60 Hz
Height (English)19.5 in.
Height (Metric)49.5 cm
Iron Total as Fe<0.5ppm
Max. TDS as CaCO3800 ppm
Particles >0.2µM/mL<1 μM/mL
Resistivity18.2MΩ·cm
Silica<30ppm
Silt Density Index<5%
Temperature Range (English)34°F to 95°F
Temperature Range (Metric)1°C to 35°C
Turbidity<1.0 NTU
Voltage100/240 VAC
Width (English)13.5 in.
Width (Metric)34.3 cm
Feed Water MonitoringNo
Feedwater SourceTap
Flow Rate12 L/hr.
Product Water TypeUltrapure (Type 1)
TOC Monitor with UV Intensity MonitoringNo
TypeReverse Osmosis System
pHOptimum Range: 6.5 to 7.5, Operating Range: 4 to 11
Unit Size-
D12661Full specifications
-24 L/hr.Request A Quote
Bacterial Content99%
Certifications/ComplianceCE, CSA
Chlorine Tolerance1000 ppm/hr.
Contaminant Rejection (Microbiological)99%
Contaminant Rejection (Minimum Ionic)Monovalent: 90 to 95%; Polyvalent: 95 to 99
Contaminant Rejection (Organic)99%
Contaminant Rejection (Particulate)90 to 95%
Depth (English)16.9 in.
Depth (Metric)42.9 cm
Electrical Requirements100/240 VAC 50/60 Hz
Feedwater Pressure30 to 100 psig (2 to 6.9 bar)
Frequency50/60 Hz
Height (English)19.5 in.
Height (Metric)49.5 cm
Iron Total as Fe<0.5ppm
Max. TDS as CaCO3800 ppm
Particles >0.2µM/mL<1 μM/mL
Resistivity18.2MΩ·cm
Silica<30ppm
Silt Density Index<5%
Temperature Range (English)34°F to 95°F
Temperature Range (Metric)1°C to 35°C
Turbidity<1.0 NTU
Voltage100/240 VAC
Width (English)13.5 in.
Width (Metric)34.3 cm
Feed Water MonitoringNo
Feedwater SourceTap
Flow Rate24 L/hr.
Product Water TypeUltrapure (Type 1)
TOC Monitor with UV Intensity MonitoringNo
TypeReverse Osmosis System
pHOptimum Range: 6.5 to 7.5, Operating Range: 4 to 11
Unit Size-
Showing 3 of 3

Intelligent Controls:

  • Automate operation, monitor system performance and pressure
  • Display key data including membrane performance, reservoir level and cartridge status
  • Monitor incoming water and membrane pressure and indicate when prefilter must be changed

Simple Operation and Maintenance:

  • Delivers multistep purification in a single system
  • Includes all required components

Technology Innovations:

  • Composite polyamide RO membranes
  • Unique MPS that protects membranes from surface scale buildup, eliminates need for softening and extends membrane life
  • Automatic flush that cleans RO membrane(s) to help eliminate buildup of scale on membrane and extends membrane life

Compact, Flexible and Quiet:

  • Small footprint fits any size lab and features convenient wall- or bench-mounting.
  • Wall bracket is included with system.
  • Optional Remote Mounting Control Panel enables placement up to 10' (3m) away.
  • 12 and 24L/hr. systems include silent-running pump.
  • 6L/hr. system uses available in-house line pressure.

Includes:

  • Prefilter
  • MPS cartridge
  • Extruded carbon filter
  • RO membrane (two supplied with D12661)
  • Wall bracket

Recommended for:

  • Buffer/reagent prep
  • Glassware washing
  • Type 1 water system feed
  • General lab use

Ordering Information:

Optional accessories include Remote Control Mounting Panel (AY1265X1) and Hot/Cold Mixing Valve (D7427). These products and replacement consumables are sold separately; contact your Sales Representative for details.

Requires:

Tank, sold separately. Contact your sales representative for details.

Figures

Frequently asked questions (FAQs)

Citations & References