Dextran, Tetramethylrhodamine, 70,000 MW, Lysine Fixable
Invitrogen™

Dextran, Tetramethylrhodamine, 70,000 MW, Lysine Fixable

Labeled dextrans are hydrophilic polysaccharides most commonly used in microscopy studies to monitor cell division, track the movement of liveRead more
Have Questions?
Catalog NumberQuantity
D181825 mg
Catalog number D1818
Price (USD)
327.18
Special offer
Online exclusive
Ends: 31-Dec-2026
399.00
Save 71.82
Each
Quantity:
25 mg
Price (USD)
327.18
Special offer
Online exclusive
Ends: 31-Dec-2026
399.00
Save 71.82
Each

Labeled dextrans are hydrophilic polysaccharides most commonly used in microscopy studies to monitor cell division, track the movement of live cells, and to report the hydrodynamic properties of the cytoplasmic matrix. The labeled dextran is commonly introduced into the cells via microinjection.

Need a different emission spectrum or longer tracking? View our other mammalian cell tracking products.

Dextran Specifications:
• Label (Ex/Em): Tetramethylrhodamine (555/580)
• Size: 70,000 MW
• Charge: Anionic
• Fixable: Fixable via Lysine

High Manufacturing Standards of Molecular Probes™ Dextrans
We offer more than 50 fluorescent and biotinylated dextran conjugates in several molecular weight ranges. Dextrans are hydrophilic polysaccharides characterized by their moderate-to-high molecular weight, good water solubility, and low toxicity. They also generally exhibit low immunogeniticy. Dextrans are biologically inert due to their uncommon poly-(α-D-1,6-glucose) linkages, which render them resistant to cleavage by most endogenous cellular glycosidases.

In most cases, Molecular Probes™ fluorescent dextrans are much brighter and have higher negative charge than dextrans available from other sources. Furthermore, we use rigorous methods for removing as much unconjugated dye as practical, and then assay our dextran conjugates by thin-layer chromatography to help ensure the absence of low molecular weight contaminants.

A Wide Selection of Substituents and Molecular Weights
Molecular Probes™ dextrans are conjugated to biotin or a wide variety of fluorophores, including seven of our Alexa Fluor™ dyes (Molecular Probes dextran conjugates–Table 14.4) and are available in these nominal molecular weights (MW): 3,000; 10,000; 40,000; 70,000; 500,000; and 2,000,000 daltons.

Dextran Net Charge and Fixability
We employ succinimidyl coupling of our dyes to the dextran molecule, which, in most cases, results in a neutral or anionic dextran. The reaction used to produce the Rhodamine Green™ and Alexa Fluor™ 488 dextrans results in the final product being neutral, anionic, or cationic. The Alexa Fluor™, Cascade Blue™, lucifer yellow, fluorescein, and Oregon Green™ dextrans are intrinsically anionic, whereas most of the dextrans labeled with the zwitterionic rhodamine B, tetramethylrhodamine, and Texas Red™ dyes are essentially neutral. To produce more highly anionic dextrans, we have developed a proprietary procedure for adding negatively charged groups to the dextran carriers; these products are designated “polyanionic” dextrans.

Some applications require that the dextran tracer be treated with formaldehyde or glutaraldehyde for subsequent analysis. For these applications, we offer “lysine-fixable” versions of most of our dextran conjugates of fluorophores or biotin. These dextrans have covalently bound lysine residues that permit dextran tracers to be conjugated to surrounding biomolecules by aldehyde-mediated fixation for subsequent detection by immunohistochemical and ultrastructural techniques. We have also shown that all of our 10,000 MW Alexa Fluor™ dextran conjugates can be fixed with aldehyde-based fixatives.

Key Applications Using Labeled Dextrans
There are a multitude of citations describing the use of labeled dextrans. Some of the most common uses include:
• Neuronal tracing (anterograde and retrograde) in live cells
• Cell lineage tracing in live cells
• Neuroanatomical tracing
• Examining intercellular communications (e.g., in gap junctions, during wound healing, and during embryonic development)
• Investigating vascular permeability and blood–brain barrier integrity
• Tracking endocytosis
• Monitoring acidification (some dextran–dye conjugates are pH-sensitive)
• Studying the hydrodynamic properties of the cytoplasmic matrix

Dextran conjugates are generally soluble in aqueous buffers to at least 10 mg/mL. Their solubilities decrease as the molecular weight increases; thus the maximum solubility in aqueous buffer is about 100 mg/mL for 3,000 MW dextrans, 50 mg/mL for 10,000 MW dextrans, 25 mg/mL for 70,000 MW dextrans and 5–10 mg/mL for the 500,000 and 2,000,000 MW dextrans.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Label or DyeClassic Dyes
Product TypeDextran
Quantity25 mg
Shipping ConditionRoom Temperature
Excitation/Emission555/580 nm
Product LineInvitrogen
Unit SizeEach
Contents & Storage
Store in freezer (-5°C to -30°C) and protect from light.

Citations & References (19)

Citations & References
Abstract
Controlled mixing in microfluidic systems using bacterial chemotaxis.
Authors:Kim MJ, Breuer KS
Journal:Anal Chem
PubMed ID:17263321
We demonstrate the use of Escherichia coli and their chemotactic characteristics to enhance mixing in a microchannel in a controlled and bi-directional manner. The presence of a chemoattractant in one arm of a three-junction microchannel results in an asymmetric increase in the effective diffusion coefficient of extremely high molecular weight ... More
Selective release of molecules from Weibel-Palade bodies during a lingering kiss.
Authors:Babich V, Meli A, Knipe L, Dempster JE, Skehel P, Hannah MJ, Carter T,
Journal:Blood
PubMed ID:18252862
Exocytosis of specialized endothelial cell secretory organelles, Weibel-Palade bodies (WPBs), is thought to play an important role in regulating hemostasis and intravascular inflammation. The major WPB core proteins are Von Willebrand factor (VWF) and its propolypeptide (Proregion), constituting more than 95% of the content. Although the composition of the WPBs ... More
The Rab5 activator ALS2/alsin acts as a novel Rac1 effector through Rac1-activated endocytosis.
Authors:Kunita R, Otomo A, Mizumura H, Suzuki-Utsunomiya K, Hadano S, Ikeda JE
Journal:J Biol Chem
PubMed ID:17409386
Mutations in the ALS2 gene cause a number of recessive motor neuron diseases, indicating that the ALS2 protein (ALS2/alsin) is vital for motor neurons. ALS2 acts as a guanine nucleotide exchange factor (GEF) for Rab5 (Rab5GEF) and is involved in endosome dynamics. However, the spatiotemporal regulation of the ALS2-mediated Rab5 ... More
Overexpression of the myelin proteolipid protein leads to accumulation of cholesterol and proteolipid protein in endosomes/lysosomes: implications for Pelizaeus-Merzbacher disease.
Authors:Simons M, Kramer EM, Macchi P, Rathke-Hartlieb S, Trotter J, Nave KA, Schulz JB
Journal:J Cell Biol
PubMed ID:11956232
Duplications and overexpression of the proteolipid protein (PLP) gene are known to cause the dysmyelinating disorder Pelizaeus-Merzbacher disease (PMD). To understand the cellular response to overexpressed PLP in PMD, we have overexpressed PLP in BHK cells and primary cultures of oligodendrocytes with the Semliki Forest virus expression system. Overexpressed PLP ... More
MLN64 mediates mobilization of lysosomal cholesterol to steroidogenic mitochondria.
Authors:Zhang M, Liu P, Dwyer NK, Christenson LK, Fujimoto T, Martinez F, Comly M, Hanover JA, Blanchette-Mackie EJ, Strauss JF
Journal:J Biol Chem
PubMed ID:12070139
This study demonstrates that the steroidogenic acute regulatory protein-related lipid transfer (START) domain-containing protein, MLN64, participates in intracellular cholesterol trafficking. Analysis of the intracellular itinerary of MLN64 and MLN64 mutants tagged with green fluorescent protein showed that the N-terminal transmembrane domains mediate endocytosis of MLN64 from the plasma membrane to ... More